iCAP™ TQe ICP-MS
iCAP™ TQe ICP-MS
iCAP™ TQe ICP-MS
iCAP™ TQe ICP-MS
iCAP™ TQe ICP-MS
Thermo Scientific™
iCAP™ TQe ICP-MS
Discover how the Thermo Scientific™ iCAP™ TQe triple quadrupole ICP-MS makes trace elemental analysis easier in applied analytical science, by avoiding the interferences that can cause sample failures and featuring improved detection limits and increased confidence in data quality.
Have Questions?
Catalog NumberType
BRE731432Triple Quadrupole ICP-MS
Catalog number BRE731432
Price (MXN)
-
Type:
Triple Quadrupole ICP-MS
Discover superior interference removal and a simpler path to accurate results with the Thermo Scientific™ iCAP™ TQe ICP-MS. This triple quadrupole ICP-MS system is the perfect tool for laboratories in applied analytical science, which require increasingly reduced detection limits, larger sample loads and greater confidence in data quality from a single sample run. The iCAP TQe ICP-MS offers a unique combination of capabilities, combining the advantages of a true triple quadrupole setup with a simple and straightforward approach for interference removal.
Why triple quadrupole ICP-MS?
Triple quadrupole ICP-MS systems offer:
  • Superior interference removal based on selective reaction chemistry in the collision/reaction cell (CRC)
  • A wide variety of interferences can be removed with a powerful combination of only two CRC gases
  • Effective suppression of unwanted side reactions when combined with a mass filtration step before the CRC
What challenges can a triple quadrupole ICP-MS resolve?
Triple quadrupole ICP-MS systems can:
  • Overcome interferences that causing unexpected sample failures, for example on As, Se, Cd and Hg
  • Improve detection limits for a wide variety of common analytes
  • Increase confidence in data quality, regardless of sample type
Why choose the iCAP TQe ICP-MS?
The iCAP TQe ICP-MS is:
  • The smallest triple quadrupole instrument on the market, requiring less bench space than other systems
  • Easy to use and maintain
  • Designed for straightforward method developement using the Reaction Finder Method Development Assistant
Specifications
DescriptioniCAP TQe ICP-MS
TypeTriple Quadrupole ICP-MS
Unit SizeEach