2-Methoxyethanol, LabServ Pronalys™
LabServ Pronalys™

2-Methoxyethanol, LabServ Pronalys™

2-Methoxyethanol Certified AR 20L
Have Questions?
Catalog number BSPM-4101-20
also known as BSPM-4101-20L
Price (USD)
-
2-Methoxyethanol Certified AR 20L
  • 2-Methoxyethanol Certified AR 20L Ethylene glycol monomethyl ether, Methyl Glycol Plastic Bottle Cas No. 109-86-4
Specifications
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
Melting Point-85°C
Molecular FormulaC3H8O2
PackagingPlastic
SMILESCOCCO
Boiling Point124°C to 125°C
GradeAnalytical
Molecular Weight (g/mol)76.10
Quantity20 L
Unit SizeEach