Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
Fisher Chemical™

Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™

Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
500 g
Packaging:
Plastic powder jar
產品號碼 C-1500-50
亦稱為 C/1500/50
價格 (TWD)
-
Quantity:
250 g
Packaging:
Plastic powder jar
Request bulk or custom format
規格
CAS10035-04-8
ColorBeige to White
Formula Weight147.02g/mol
IUPAC Namecalcium dihydrate dichloride
InChI KeyLLSDKQJKOVVTOJ-UHFFFAOYSA-L
檢視更多
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP