What types of large vessel endothelial cells do you offer?
We carry primary human umbilical vein endothelial cells (HUVEC) from one donor or pooled from multiple donors.
Search
View additional product information for Human Umbilical Vein Endothelial Cells, HUVEC Pooled, (1 x 106 cells) - FAQs (C01510C)
1 product FAQs found