Thermo Scientific™
Rocket Synergy™ 2 Evaporator Preventative Maintenance and Service Kits
Catalog number: C0960-01-00094
Thermo Scientific™
Rocket Synergy™ 2 Evaporator Preventative Maintenance and Service Kits
Catalog number: C0960-01-00094
Choose Thermo Scientific™ Rocket Synergy™ 2 Evaporator Preventative Maintenance and Service Kits to keep your Rocket Synergy 2 Evaporator running smoothly and to avoid down time.
Catalog Number
C0960-01-00094
Unit Size
Each
Type
Preventative Maintenance Kit
For Use With (Equipment)
Rocket Synergy™ 2 IGP System
Price (USD)
Full specifications
DescriptionRocket Synergy™ 2 IGP System Preventative Maintenance Kit
TypePreventative Maintenance Kit
For Use With (Equipment)Rocket Synergy™ 2 IGP System
Quantity1 Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Preventative Maintenance Service Kit
For Use With (Equipment)
Rocket Synergy™ 2 Evaporator Rotor
Price (USD)
Full specifications
DescriptionRocket Synergy™ 2 Evaporator Preventative Maintenance Service Kit, Rotor
TypePreventative Maintenance Service Kit
For Use With (Equipment)Rocket Synergy™ 2 Evaporator Rotor
Quantity1 Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Preventative Maintenance Kit
For Use With (Equipment)
Rocket Synergy™ 2 System
Price (USD)
Full specifications
DescriptionRocket Synergy™ 2 System Preventative Maintenance Kit
TypePreventative Maintenance Kit
For Use With (Equipment)Rocket Synergy™ 2 System
Quantity1 Kit
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeTypeFor Use With (Equipment)Price (USD)
C0960-01-00094Full specifications
EachPreventative Maintenance KitRocket Synergy™ 2 IGP SystemRequest A Quote
DescriptionRocket Synergy™ 2 IGP System Preventative Maintenance Kit
TypePreventative Maintenance Kit
For Use With (Equipment)Rocket Synergy™ 2 IGP System
Quantity1 Kit
Unit SizeEach
00960-01-00058Full specifications
EachPreventative Maintenance Service KitRocket Synergy™ 2 Evaporator RotorRequest A Quote
DescriptionRocket Synergy™ 2 Evaporator Preventative Maintenance Service Kit, Rotor
TypePreventative Maintenance Service Kit
For Use With (Equipment)Rocket Synergy™ 2 Evaporator Rotor
Quantity1 Kit
Unit SizeEach
C0960-01-00093Full specifications
EachPreventative Maintenance KitRocket Synergy™ 2 SystemRequest A Quote
DescriptionRocket Synergy™ 2 System Preventative Maintenance Kit
TypePreventative Maintenance Kit
For Use With (Equipment)Rocket Synergy™ 2 System
Quantity1 Kit
Unit SizeEach
Showing 3 of 3
Spend minimal time and effort to concentrate or dry large-volume samples on the Rocket Synergy 2 Evaporator for chromatographic analysis. This system provides unattended sample evaporation and with excellent reproducibility and recovery rates. Choose from several sample formats including:
  • Thermo Scientific™ Dionex™ ASE™ Accelerated Solvent Extractor vials for convenient workflow from the Dionex ASE system to the Rocket Synergy 2 Evaporator
  • 450 mL flasks for large-volume samples
  • Thermo Scientific™ Rocket™ Synergy 2 Evaporator SampleGenie™ vials and SampleGenie for GC autosampler vials for smaller volume samples
  • Rocket Synergy 2 Evaporator Flip-Flop vials for collecting samples in extraction vials and transferring and evaporating them directly into GC vials