Click-iT™ Protein Reaction Buffer Kit - FAQs
Click-iT™ Protein Reaction Buffer Kit - FAQs
View additional product information for Click-iT™ Protein Reaction Buffer Kit - FAQs (C10276)
1 product FAQs found
What are the main characteristics of a Click-iT reaction?
Click reactions have several general characteristics: the reaction is efficient, no extreme temperatures or solvents are required, the reaction is complete within 30 minutes, the components of the reaction are bio-inert, and perhaps most importantly, no side reactions occur-the label and detection tags react selectively and specifically with one another. This final point is a key advantage of this powerful detection technique; it is possible to apply click chemistry-labeled molecules to complex biological samples and detect them with unprecedented sensitivity due to the extremely low background of the reaction.