Click-iT™ EdU Alexa Fluor™ 488 HCS Assay
Invitrogen™

Click-iT™ EdU Alexa Fluor™ 488 HCS Assay

The Click-iT™ EdU Alexa Fluor™ 488 HCS Assay is a superior alternative to traditional proliferation assays that is optimized for深入閱讀
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
C103502 x 96 well plates
C1035110 x 96 well plates
產品號碼 C10350
價格 (TWD)
26,600.00
線上優惠
Ends: 31-Dec-2026
38,000.00
您節省 11,400.00 (30%)
Each
Quantity:
2 x 96 well plates
價格 (TWD)
26,600.00
線上優惠
Ends: 31-Dec-2026
38,000.00
您節省 11,400.00 (30%)
Each
The Click-iT™ EdU Alexa Fluor™ 488 HCS Assay is a superior alternative to traditional proliferation assays that is optimized for high-content imaging applications. In this assay the modified thymidine analogue EdU is efficiently incorporated into newly synthesized DNA and fluorescently labeled with a bright, photostable Alexa Fluor™ dye in a fast, highly-specific click reaction. This fluorescent labeling of proliferating cells is accurate and compatible with antibody methods due to the mild click protocol.

The Click-iT™ EdU Alexa Fluor™ 488 HCS Assay is:

• Simple—works the first time, every time, in less time
• Efficient—no denaturation steps or harsh treatment required
• Gentle on samples—better preservation of cell morphology, antigen structure, and DNA integrity
• Consistent—not dependent on variable antibody lots for detection

The most accurate proliferation-detection methods are based on the incorporation and measurement of nucleoside analogues in newly synthesized DNA, with bromodeoxyuridine (BrdU) a commonly used analogue. BrdU-labeled DNA is quantitated using anti-BrdU antibodies following DNA denaturation by harsh methods (HCl, heat, or enzymes) to expose the BrdU molecules. This step is time consuming and difficult to perform consistently. The harsh treatment can also adversely affect sample integrity and quality, which makes co-staining with other antibodies challenging.

Superior Proliferation Methodology
The Click-iT™ EdU Alexa Fluor™ 488 HCS Assay provides a superior alternative to BrdU assays for measuring cell proliferation. EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog of thymidine and is incorporated into DNA during active DNA synthesis. With Click-iT™ EdU, mild fixation and detergent permeabilization is sufficient for the small molecule-based Click-iT™ EdU detection reagent to gain access to the DNA. As a consequence, the Click-iT™ EdU HCS assay is not only easy to use, but more accurate and compatible with cell cycle analysis and other intracellular or extracellular targets for truly content-rich results.

The kit is optimized for high-content imaging applications; visit the Click-iT™ technology area of our website for kits designed for fluorescence microscopy, microplate, or flow cytometry platforms.

Learn more about Click-iT™ technology

Notes:
The Click-iT™ assay can be used on cells in culture or in vivo following administration of EdU by feeding or injection methods.
The Click-iT™ assay can be used with BrdU in dual pulse experiments by using the ant-BrdU (clone MoBu-1) antibody, which does not cross react with EdU.
The Click-iT™ technology is compatible with immunohistochemical, immunocytochemical, and fluorescent dyes that are fixation tolerant or designed for fixed-cell labeling.
For Research Use Only. Not for use in diagnostic procedures.
規格
Detection MethodFluorescence
Dye TypeAlexa Fluor™ 488
Emission WavelengthVisible
Format96-well plate
Quantity2 x 96 well plates
Shipping ConditionRoom Temperature
ColorGreen
For Use With (Application)High-content Screening (HCS)
For Use With (Equipment)High Content Analysis Instrument
Product LineAlexa Fluor, Click-iT, Molecular Probes
Product TypeHCS Assay
Unit SizeEach
內容物與存放
The kit contains sufficient material for 2 96-well plates. Store at 2–6°C, dessicate and protect from light.

常見問答集 (常見問題)

What are the main characteristics of a Click-iT reaction?

Click reactions have several general characteristics: the reaction is efficient, no extreme temperatures or solvents are required, the reaction is complete within 30 minutes, the components of the reaction are bio-inert, and perhaps most importantly, no side reactions occur-the label and detection tags react selectively and specifically with one another. This final point is a key advantage of this powerful detection technique; it is possible to apply click chemistry-labeled molecules to complex biological samples and detect them with unprecedented sensitivity due to the extremely low background of the reaction.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I will be performing a cell proliferation assay using Click-iT EdU kit. At what point can I stop overnight, or do I have to perform all the steps continuously?

One may store the sample after fixation overnight in PBS at 4oC. For longer storage (<1 week) , store in buffer with 1-2% formaldehyde or in formalin to limit microbial growth. If you use sodium azide as a microbial inhibitor, it must be completely removed prior to the Click-iT reaction.

Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center.

A control for a Click-iT EdU labeling experiment uses no EdU and the Click-iT reaction using Alexa Fluor 594 azide. The mouse heart tissue sections are showing non-specific labeling in red, seen in particular clusters of cells. They don't overlap with DAPI. What is the problem?

The problem is likely not the Alexa Fluor 594 azide. Since there are no alkynes endogenous to mouse tissue, there is nothing for the dye-azide to bind to. Since the background doesn't overlap with nuclei (DAPI signal), this isn't an issue of unintended EdU labeling. This red is autofluorescence from red blood cells; they autofluoresce in the red and don't have nuclei. This can be confirmed by checking a completely unlabeled tissue section (no dye present at all) to see if they are still present and by examining the cells at high magnification and looking for corpuscular shape.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the Alexa Fluor azides from Click-iT EdU kits available separately?

Yes, but the standalone products are not shipped at the same amount as provided in the Click-iT EdU kits; the amount of dye-azide provided in the Click-iT kits is proprietary information. See these catalog numbers for the stand alone products:
- Cat. No. A10266: Alexa Fluor 488 azide
- Cat. No. A10270: Alexa Fluor 594 azide
- Cat. No. A10277: Alexa Fluor 647 azide

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am observing no signal or very low specific signal for my click-labeled samples. What can I do to improve the signal?

The click reaction is only effective when copper is in the appropriate valency. Azides and alkynes will not react with each other without copper. Make sure that the click reaction mixture is used immediately after preparation when the copper (II) concentration is at its highest.
Do not use additive buffer that has turned yellow; it must be colorless to be active.
Cells need to be adequately fixed and permeabilized for the TdT enzyme and click reagents to have access to the nucleus. Tissue samples require digestion with proteinase K or other proteolytic enzymes for sufficient TdT access.
Some reagents can bind copper and reduce its effective concentration available to catalyze the click reaction. Do not include any metal chelator (e.g., EDTA, EGTA, citrate, etc.) in any buffer or reagent prior to the click reaction. Avoid buffers or reagents that include other metal ions that may be o xidized or reduced. It may be help to include extra wash steps on the cell or tissue sample before performing the click reaction.
You can repeat the click reaction with fresh reagents to try to improve signal. Increasing the click reaction time longer than 30 minutes will not improve a low signal. Performing a second, 30 minute incubation with fresh click reaction reagents is more effective at improving labeling.
Your cells may not be apoptotic. Prepare a DNase I-treated positive control to verify that the TdT enzymatic reaction and click labeling reaction are working correctly.

Find additional tips, troubleshooting help, and resources within our Labeling Chemistry Support Center.

引用資料與參考文獻 (12)

引用資料與參考文獻
Abstract
Cell type specific applicability of 5-ethynyl-2'-deoxyuridine (EdU) for dynamic proliferation assessment in flow cytometry.
Authors:Diermeier-Daucher S, Clarke ST, Hill D, Vollmann-Zwerenz A, Bradford JA, Brockhoff G,
Journal:Cytometry A
PubMed ID:19235202
'Using the nucleoside analogue EdU (5-ethynyl-2''-deoxyuridine) for thymidine substitution instead of BrdU (5-bromo-2''-deoxyuridine) in cell proliferation assays has recently been proposed. However, the effect of EdU on cell viability, DNA synthesis, and cell cycle progression and consequently its usability for dynamic cell proliferation analysis in vitro has not been explored. ... More
Imaging and analysis of 3D tumor spheroids enriched for a cancer stem cell phenotype.
Authors:Robertson FM, Ogasawara MA, Ye Z, Chu K, Pickei R, Debeb BG, Woodward WA, Hittelman WN, Cristofanilli M, Barsky SH,
Journal:J Biomol Screen
PubMed ID:20639504
'Tumors that display a highly metastatic phenotype contain subpopulations of cells that display characteristics similar to embryonic stem cells. These cells exhibit the ability to undergo self-renewal; slowly replicate to retain a nucleoside analog label, leading to their definition as ' ... More
Development of a high-content screening assay panel to accelerate mechanism of action studies for oncology research.
Authors:Towne DL, Nicholl EE, Comess KM, Galasinski SC, Hajduk PJ, Abraham VC,
Journal:J Biomol Screen
PubMed ID:22706350
'Efficient elucidation of the biological mechanism of action of novel compounds remains a major bottleneck in the drug discovery process. To address this need in the area of oncology, we report the development of a multiparametric high-content screening assay panel at the level of single cells to dramatically accelerate understanding ... More
EdU, a new thymidine analogue for labelling proliferating cells in the nervous system.
Authors:Chehrehasa F, Meedeniya AC, Dwyer P, Abrahamsen G, Mackay-Sim A,
Journal:J Neurosci Methods
PubMed ID:18996411
Labelling and identifying proliferating cells is central to understanding neurogenesis and neural lineages in vivo and in vitro. We present here a novel thymidine analogue, ethynyl deoxyuridine (EdU) for labelling dividing cells, detected with a fluorescent azide which forms a covalent bond via the  ... More
Genome-wide RNAi screen identifies genes involved in intestinal pathogenic bacterial infection.
Authors:Cronin SJ, Nehme NT, Limmer S, Liegeois S, Pospisilik JA, Schramek D, Leibbrandt A, Simoes Rde M, Gruber S, Puc U, Ebersberger I, Zoranovic T, Neely GG, von Haeseler A, Ferrandon D, Penninger JM,
Journal:Science
PubMed ID:19520911
Innate immunity represents the first line of defense in animals. We report a genome-wide in vivo Drosophila RNA interference screen to uncover genes involved in susceptibility or resistance to intestinal infection with the bacterium Serratia marcescens. We first employed whole-organism gene suppression, followed by tissue-specific silencing in gut epithelium or ... More