Click-iT™ EdU Pacific Blue™ Flow Cytometry Assay Kit
Invitrogen™

Click-iT™ EdU Pacific Blue™ Flow Cytometry Assay Kit

The Click-iT™ EdU Pacific Blue™ Flow Cytometry Assay Kit provides a simplified, more robust assay for analyzing DNA replication inRead more
Have Questions?
Catalog NumberQuantity
C1041850 Assays
Catalog number C10418
Price (USD)
788.00
Each
Quantity:
50 Assays
Price (USD)
788.00
Each
The Click-iT™ EdU Pacific Blue™ Flow Cytometry Assay Kit provides a simplified, more robust assay for analyzing DNA replication in proliferating cells as compared to traditional BrdU methods. Newly synthesized DNA is analyzed using the 405 nm laser of the flow cytometer.

• Accurate—superior results compared to BrdU assays
• Fast—results in as little as 90 minutes
• Economical—more assays per kit

View selection guide for all Click-iT™ EdU and Click-iT™ Plus EdU assays for flow cytometry.

An Advanced Method Giving You Results Superior to BrdU
The most accurate method of proliferation analysis is direct measurement of DNA synthesis. Originally, this was performed through incorporation of radioactive nucleosides, i.e., 3H-thymidine. This method was replaced by antibody-based detection of the nucleoside analog bromodeoxyuridine (BrdU). The Click-iT™ EdU Flow Cytometry Assay Kits are novel alternatives to the BrdU assay. EdU (5-ethynyl-2´-deoxyuridine) is a thymidine analog which is incorporated into DNA during active DNA synthesis. Detection is based on click chemistry: a copper catalyzed covalent reaction between an azide and an alkyne. In this application, the alkyne is found in the ethynyl moiety of EdU, while the azide is coupled to Alexa Fluor ™ 488, Alexa Fluor™ 647, or Pacific Blue™ dyes. Standard flow cytometry methods are used for determining the percentage of S-phase cells in the population (Fig 1).

Mild Conditions Allow Use with Cell Cycle Dyes and Antibodies
The advantages of Click-iT™ EdU labeling are readily evident while performing the assay (Fig 2). The small size of the dye azide allows for efficient detection of the incorporated EdU using mild conditions, while standard aldehyde-based fixation and detergent permeabilization are sufficient for the Click-iT™ detection reagent to gain access to the DNA. This is in contrast to BrdU assays that require DNA denaturation (using HCl, heat, or digestion with DNase) to expose the BrdU so that it may be detected with an anti-BrdU antibody. Sample processing for the BrdU assay can result in signal alteration of the cell cycle distribution as well as destruction of antigen recognition sites when using the HCl method. In contrast, the easy-to-use EdU cell proliferation kit is compatible with cell cycle dyes. This EdU assay kit can also be multiplexed with antibodies against surface and intracellular markers.

Quick and Simple Protocol
The Click-iT™ EdU protocol is based on the aldehyde fixation and detergent permeabilization steps for immunohistochemical antibody labeling, but EdU is compatible with other fixation/permeabilization agents including saponin and methanol. In just five steps you’ll be ready to analyze your cell proliferation data:

• Treat cells with EdUM
• Fix and permeabilize cells
• Detect S-phase cells with Click-iT™ detection cocktail for 30 min
• Wash once
• Analyze

Get Accurate Results Economically
By increasing the number of assays per kit, the Click-IT™ EdU Pacific Blue™ Flow Cytometry Assay Kit is less expensive than the traditional BrdU assays making them ideal for large experiments.

For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypePacific Blue™
Emission Wavelength404/450
FormatTube(s)
Quantity50 Assays
Shipping ConditionRoom Temperature
For Use With (Equipment)Flow Cytometer
Product LineClick-iT, Pacific Blue
Product TypeFlow Cytometry Assay Kit
Unit SizeEach
Contents & Storage
Contains EdU (5-ethynyl-2' -deoxyuridine), Pacific Blue™ azide, anhydrous dimethylsulfoxide (DMSO), Click-iT™ fixative, Click-iT™ saponin-based permeabilization and wash buffer, copper (II) sulfate,, and Click-iT™ EdU buffer additive.
  • Store at 2°C to 8°C
  • Dessicate and protect from light.

    Frequently asked questions (FAQs)

    What are the main characteristics of a Click-iT reaction?

    Click reactions have several general characteristics: the reaction is efficient, no extreme temperatures or solvents are required, the reaction is complete within 30 minutes, the components of the reaction are bio-inert, and perhaps most importantly, no side reactions occur-the label and detection tags react selectively and specifically with one another. This final point is a key advantage of this powerful detection technique; it is possible to apply click chemistry-labeled molecules to complex biological samples and detect them with unprecedented sensitivity due to the extremely low background of the reaction.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    I will be performing a cell proliferation assay using Click-iT EdU kit. At what point can I stop overnight, or do I have to perform all the steps continuously?

    One may store the sample after fixation overnight in PBS at 4oC. For longer storage (<1 week) , store in buffer with 1-2% formaldehyde or in formalin to limit microbial growth. If you use sodium azide as a microbial inhibitor, it must be completely removed prior to the Click-iT reaction.

    Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center.

    Can I combine Click-iT or Click-iT Plus reactions with phalloidin conjugates used for actin staining?

    We do not recommend using phalloidin conjugates for staining actin in combination with traditional Click-iT or Click-iT Plus reactions since phalloidin is extremely sensitive to the presence of copper.

    For staining actin in combination with traditional Click-iT or Click-iT Plus reactions, we recommend using anti-α-actin antibodies for staining actin in the cytoskeleton. You can find a list of our actin antibodies here.

    Another option would be to use the Click-iT Plus Alexa Fluor Picolyl Azide Toolkit (Cat. Nos. C10641, C10642, C10643). These Click-iT Plus toolkits provide Copper and Copper protectant separately which makes it easier to titrate the copper concentration to obtain optimal labeling with minimal copper-mediated damage. You may need to optimize the click reaction with the lowest possible concentration of copper and then perform the phalloidin staining.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    I am observing no signal or very low specific signal for my click-labeled samples. What can I do to improve the signal?

    The click reaction is only effective when copper is in the appropriate valency. Azides and alkynes will not react with each other without copper. Make sure that the click reaction mixture is used immediately after preparation when the copper (II) concentration is at its highest.
    Do not use additive buffer that has turned yellow; it must be colorless to be active.
    Cells need to be adequately fixed and permeabilized for the TdT enzyme and click reagents to have access to the nucleus. Tissue samples require digestion with proteinase K or other proteolytic enzymes for sufficient TdT access.
    Some reagents can bind copper and reduce its effective concentration available to catalyze the click reaction. Do not include any metal chelator (e.g., EDTA, EGTA, citrate, etc.) in any buffer or reagent prior to the click reaction. Avoid buffers or reagents that include other metal ions that may be o xidized or reduced. It may be help to include extra wash steps on the cell or tissue sample before performing the click reaction.
    You can repeat the click reaction with fresh reagents to try to improve signal. Increasing the click reaction time longer than 30 minutes will not improve a low signal. Performing a second, 30 minute incubation with fresh click reaction reagents is more effective at improving labeling.
    Your cells may not be apoptotic. Prepare a DNase I-treated positive control to verify that the TdT enzymatic reaction and click labeling reaction are working correctly.

    Find additional tips, troubleshooting help, and resources within our Labeling Chemistry Support Center.

    I am observing high non-specific background when I image my Click-iT EdU TUNEL-labeled samples. What is causing this and what can I do to reduce the background?

    The click reaction is very selective between an azide and alkyne. No other side reactions are possible in a biological system. Any non-specific background is due to non-covalent binding of the dye to various cellular components. The Select FX Signal Enhancer is not effective at reducing non-specific charge-based binding of dyes following the click reaction; we do not recommend its use with the Click-iT detection reagents. The best method to reduce background is to increase the number of BSA washes. You should always do a no-dye or no-click reaction control under the same processing and detection conditions to verify that the background is actually due to the dye and not autofluorescence. You should also perform the complete click reaction on a no-TdT enzyme control sample to verify the specificity of the click reaction signal.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    Citations & References (5)

    Citations & References
    Abstract
    Accumulation of CD11b+ lung dendritic cells in response to fungal infection results from the CCR2-mediated recruitment and differentiation of Ly-6Chigh monocytes.
    Authors:Osterholzer JJ, Chen GH, Olszewski MA, Curtis JL, Huffnagle GB, Toews GB,
    Journal:J Immunol
    PubMed ID:19933856
    'Pulmonary clearance of the encapsulated yeast Cryptococcus neoformans is associated with the CCR2-mediated accumulation of lung dendritic cells (DC) and the development of a T1 adaptive immune response. The objective of this study was to identify the circulating DC precursor(s) responsible for this large increase in lung DC numbers. An ... More
    Enhanced expression of fibroblast growth factor receptor 2 IIIc promotes human pancreatic cancer cell proliferation.
    Authors:Ishiwata T, Matsuda Y, Yamamoto T, Uchida E, Korc M, Naito Z,
    Journal:Am J Pathol
    PubMed ID:22440254
    'In pancreatic ductal adenocarcinoma (PDAC), the fibroblast growth factor receptor 1 (FGFR-1) IIIb isoform correlates with the inhibition of cancer cell proliferation, migration, and invasion, whereas FGFR-1 IIIc enhances cancer cell proliferation. The FGFR-2 IIIb isoform is expressed in PDAC, and its expression correlates with increased venous invasion. We examined ... More
    Loss of epidermal Evi/Wls results in a phenotype resembling psoriasiform dermatitis.
    Authors:Augustin I, Gross J, Baumann D, Korn C, Kerr G, Grigoryan T, Mauch C, Birchmeier W, Boutros M,
    Journal:
    PubMed ID:23918954
    Cells of the epidermis renew constantly from germinal layer stem cells. Although epithelial cell differentiation has been studied in great detail and the role of Wnt signaling in this process is well described, the contribution of epidermal Wnt secretion in epithelial cell homeostasis remains poorly understood. To analyze the role ... More
    IL-7 abrogates suppressive activity of human CD4+CD25+FOXP3+ regulatory T cells and allows expansion of alloreactive and autoreactive T cells.
    Authors:Heninger AK, Theil A, Wilhelm C, Petzold C, Huebel N, Kretschmer K, Bonifacio E, Monti P,
    Journal:J Immunol
    PubMed ID:23129754
    CD4(+)CD25(+)FOXP3(+) regulatory T cells (Tregs) control the activation and expansion of alloreactive and autoreactive T cell clones. Because uncontrolled activation and expansion of autoreactive T cells occur in an IL-7-rich environment, we explored the possibility that IL-7 may affect the function of Treg. We show that the functional high-affinity IL-7R ... More
    13q14 deletions in CLL involve cooperating tumor suppressors.
    Authors:Palamarchuk A, Efanov A, Nazaryan N, Santanam U, Alder H, Rassenti L, Kipps T, Croce CM, Pekarsky Y,
    Journal:Blood
    PubMed ID:20071661
    B-cell chronic lymphocytic leukemia (CLL) is the most common human leukemia. 13q14 deletions are most common chromosomal alterations in CLL. We previously reported that miR-15/16 is a target of 13q14 deletions and plays a tumor suppressor role by targeting BCL2. Because DLEU7 is located near miR-15/16 and is also positioned ... More