CellROX™ Reagent for oxidative stress detection
Invitrogen™

CellROX™ Reagent for oxidative stress detection

CellROX™ Reagents are novel fluorogenic probes for measuring cellular oxidative stress in both live and fixed cell imaging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
C10422Deep Red
C10448Orange, Far-Red, Green
C10444Green
C10443Orange
Catalog number C10422
Price (USD)
655.65
Online exclusive
690.00
Save 34.35
Each
Color:
Deep Red
Price (USD)
655.65
Online exclusive
690.00
Save 34.35
Each

The CellROX™ Reagents are novel fluorogenic probes for measuring cellular oxidative stress in live cells. These cell-permeant dyes exhibit bright fluorescence upon oxidation by reactive oxygen species (ROS). (See table below for details of each reagent.)

The CellROX™ reagents are:

  • Optimized for detection of oxidative stress in live cells and may be added directly to serum-containing growth media
  • Provided as a stable, ready-to-use DMSO solutions with simple protocols, compatible with standard workflows in fluorescence microscopy
  • Compatible with a variety of detection platforms, including traditional fluorescence microscopy, high content screening (HCS), and flow cytometry

Oxidative stress results from an imbalance between the production of reactive oxygen species (ROS) and the ability of cells to scavenge them. ROS play an important role in the progression of several diseases, including inflammation, atherosclerosis, aging, and age-related degenerative disorders.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorDeep Red
Concentration2.5 mM stabilized solution in DMSO
For Use With (Equipment)Imaging, HCS, Cytometer
FormatLiquid
Quantity5 x 50 μL
Detection MethodLive Cell Imaging
Excitation/Emission644/665 nm
IndicatorOxidative stress
Product LineCellROX
Unit SizeEach
Contents & Storage
Store at ≤–20°C. Protect from light and desiccate.

Frequently asked questions (FAQs)

I want to assay cells for reactive oxygen species using carboxy-H2DCFDA, but I want to do so with a plate reader instead of microscope. Will it work?

It has been done. The problem is that plate readers are less sensitive than microscopes, with far less signal-to-background difference. It is worth trying, but first optimize concentrations and loading times with control cells, use a plate with little to no autofluorescence, and possibly optimize the gain setting in order to get the best signal possible. But don't expect the same sensitivity, even with optimization.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I have GFP-transfected cells and need to label for reactive oxygen species. Can I use H2DCFDA?

This is not recommended as the two dyes overlap in the emission wavelength. There are other ROS reagents available in different wavelengths, such as CellROX Deep Red, which emits in the far-red range (665 nm), or dihydroethidium, which is emits in the visible red range (620 nm).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I need a formaldehyde-fixable reactive oxygen species detection assay. Is H2 DCFDA fixable?

H2DCFDA and similar derivatives are not fixable. The same goes for dihydroethidium and dihydrorhodamine. However, CellROX Deep Red and CellROX Green are retained for a limited time upon fixation with formaldehyde. CellROX Green may be retained upon subsequent Triton X-100 permeabilization. Avoid the use of any acetone or alcohol-based fixatives or fixatives that include alcohol, such as formalin.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What dyes can I use to detect reactive oxygen species (ROS) in my bacteria?

Many dyes that are used on mammalian cells have also been shown to be useful in bacterial cells. For example, CellROX Deep Red Reagent has been shown to work in B. subtilis (see Reference: http://www-brs.ub.ruhr-uni-bochum.de/netahtml/HSS/Diss/RaatschenNadja/diss.pdf). If you are interested in a particular dye, but are not sure if it will work on your bacteria, literature searches are the best way to check to see if it has been tested. If not, then it may be worth testing yourself.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am trying to label my cells with a reactive oxygen species (ROS) indicator dye, but I am not seeing a significant difference in signal. What could be happening?

First, make sure you have both a negative (untreated) and positive (ROS-induced) sample to compare. A good positive control can be the use of 100 µM menadione for one hour or 50 µM nefazodone for 24 hours. H2O2 can also be used, though it does not work well for CellROX dyes. Some dyes, such as H2DCFDA, require esterase cleavage, so don't incubate in the presence of serum (which contains esterases that can prematurely cleave the dye). If your positive control does not show significant change compared to the negative control, try increasing the concentration and label time for the dye. Our manuals give starting recommendations. Be sure to image your live cells as soon as possible. Only two dyes (CellROX Green and CellROX Deep Red) are retained with formaldehyde fixation. Finally, make sure you are using filters and instrument settings to match the excitation and emission spectra of the dye.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (52)

Citations & References
Abstract
The airway epithelium nucleotide-binding domain and leucine-rich repeat protein 3 inflammasome is activated by urban particulate matter.
Authors:Hirota JA, Hirota SA, Warner SM, Stefanowicz D, Shaheen F, Beck PL, Macdonald JA, Hackett TL, Sin DD, Van Eeden S, Knight DA,
Journal:J Allergy Clin Immunol
PubMed ID:22227418
The airway epithelium is the first line of defense against inhaled insults and therefore must be capable of coordinating appropriate inflammatory and immune responses. We sought to test the hypothesis that the nucleotide-binding domain and leucine-rich repeat protein 3 (NLRP3) inflammasome, an intracellular danger-sensing complex, plays a critical role in ... More
Combination Small Molecule MEK and PI3K Inhibition Enhances Uveal Melanoma Cell Death in a Mutant GNAQ- and GNA11-Dependent Manner.
Authors:Khalili JS, Yu X, Wang J, Hayes BC, Davies MA, Lizee G, Esmaeli B, Woodman SE,
Journal:Clin Cancer Res
PubMed ID:22733540
Activating Q209L/P mutations in GNAQ or GNA11 (GNAQ/11) are present in approximately 80% of uveal melanomas. Mutant GNAQ/11 are not currently therapeutically targetable. Inhibiting key down-stream effectors of GNAQ/11 represents a rational therapeutic approach for uveal melanomas that harbor these mutations. The mitogen-activated protein/extracellular signal-regulated kinase/mitogen-activated protein kinase (MEK/MAPK) and ... More
Mutant BRAF induces DNA strand breaks, activates DNA damage response pathway, and up-regulates glucose transporter-1 in nontransformed epithelial cells.
Authors:Sheu JJ, Guan B, Tsai FJ, Hsiao EY, Chen CM, Seruca R, Wang TL, Shih IeM,
Journal:Am J Pathol
PubMed ID:22227015
'Although the oncogenic functions of activating BRAF mutations have been clearly demonstrated in human cancer, their roles in nontransformed epithelial cells remain largely unclear. Investigating the cellular response to the expression of mutant BRAF in nontransformed epithelial cells is fundamental to the understanding of the roles of BRAF in cancer ... More
Mechanisms of programmed cell death signaling in hair cells and support cells post-electrode insertion trauma.
Authors:Eshraghi AA, Lang DM, Roell J, Van De Water TR, Garnham C, Rodrigues H, Guardiola M, Gupta C, Mittal J,
Journal:
PubMed ID:25761716
'Programmed cell death (PCD) initially starts in the support cells (SCs) after electrode insertion trauma (EIT), followed by PCD in hair cells (HCs). Activation of caspase-3 was observed only in SCs. Protecting both SCs and HCs with selective otoprotective drugs at an early stage post implantation may help to preserve ... More
OPA1 Mutation and Late-Onset Cardiomyopathy: Mitochondrial Dysfunction and mtDNA Instability.
Authors:Chen L, Liu T, Tran A, Lu X, Tomilov AA, Davies V, Cortopassi G, Chiamvimonvat N, Bers DM, Votruba M, Knowlton AA,
Journal:J Am Heart Assoc
PubMed ID:23316298
'Mitochondrial fusion protein mutations are a cause of inherited neuropathies such as Charcot-Marie-Tooth disease and dominant optic atrophy. Previously we reported that the fusion protein optic atrophy 1 (OPA1) is decreased in heart failure. We investigated cardiac function, mitochondrial function, and mtDNA stability in a mouse model of the disease ... More