CellROX™ Reagent for oxidative stress detection
Invitrogen™

CellROX™ Reagent for oxidative stress detection

CellROX™ Reagents are novel fluorogenic probes for measuring cellular oxidative stress in both live and fixed cell imaging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
C10443Orange
C10448Orange, Far-Red, Green
C10444Green
C10422Deep Red
Catalog number C10443
Price (USD)
562.52
Special offer
Online exclusive
Ends: 31-Dec-2026
686.00
Save 123.48
Each
Color:
Orange
Price (USD)
562.52
Special offer
Online exclusive
Ends: 31-Dec-2026
686.00
Save 123.48
Each

The CellROX™ Reagents are novel fluorogenic probes for measuring cellular oxidative stress in live cells. These cell-permeant dyes exhibit bright fluorescence upon oxidation by reactive oxygen species (ROS). (See table below for details of each reagent.)

The CellROX™ reagents are:

  • Optimized for detection of oxidative stress in live cells and may be added directly to serum-containing growth media
  • Provided as a stable, ready-to-use DMSO solutions with simple protocols, compatible with standard workflows in fluorescence microscopy
  • Compatible with a variety of detection platforms, including traditional fluorescence microscopy, high content screening (HCS), and flow cytometry

Oxidative stress results from an imbalance between the production of reactive oxygen species (ROS) and the ability of cells to scavenge them. ROS play an important role in the progression of several diseases, including inflammation, atherosclerosis, aging, and age-related degenerative disorders.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorOrange
Concentration2.5 mM stabilized solution in DMSO
For Use With (Equipment)Imaging, HCS, Cytometer
FormatFrozen
Quantity5 x 50 μL
Detection MethodLive Cell Imaging
Excitation/Emission545/565 nm
IndicatorOxidative stress
Product LineCellROX
Unit SizeEach
Contents & Storage
Store at ≤–20°C. Protect from light and desiccate.

Frequently asked questions (FAQs)

What dyes can I use to detect reactive oxygen species (ROS) in my bacteria?

Many dyes that are used on mammalian cells have also been shown to be useful in bacterial cells. For example, CellROX Deep Red Reagent has been shown to work in B. subtilis (see Reference: http://www-brs.ub.ruhr-uni-bochum.de/netahtml/HSS/Diss/RaatschenNadja/diss.pdf). If you are interested in a particular dye, but are not sure if it will work on your bacteria, literature searches are the best way to check to see if it has been tested. If not, then it may be worth testing yourself.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am trying to label my cells with a reactive oxygen species (ROS) indicator dye, but I am not seeing a significant difference in signal. What could be happening?

First, make sure you have both a negative (untreated) and positive (ROS-induced) sample to compare. A good positive control can be the use of 100 µM menadione for one hour or 50 µM nefazodone for 24 hours. H2O2 can also be used, though it does not work well for CellROX dyes. Some dyes, such as H2DCFDA, require esterase cleavage, so don't incubate in the presence of serum (which contains esterases that can prematurely cleave the dye). If your positive control does not show significant change compared to the negative control, try increasing the concentration and label time for the dye. Our manuals give starting recommendations. Be sure to image your live cells as soon as possible. Only two dyes (CellROX Green and CellROX Deep Red) are retained with formaldehyde fixation. Finally, make sure you are using filters and instrument settings to match the excitation and emission spectra of the dye.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (18)

Citations & References
Abstract
MicroRNA-27 (miR-27) Targets Prohibitin and Impairs Adipocyte Differentiation and Mitochondrial Function in Human Adipose-derived Stem Cells.
Authors:Kang T, Lu W, Xu W, Anderson L, Bacanamwo M, Thompson W, Chen YE, Liu D,
Journal:
PubMed ID:24133204
Prohibitin (PHB) has been reported to play a crucial role in adipocyte differentiation and mitochondrial function. However, the regulative mechanism of PHB during adipogenesis remains unclear. In this study, we determined that the levels of both microRNA (miR)-27a and miR-27b were down-regulated following adipogenic induction of human adipose-derived stem cells, ... More
Mitohormesis in Hypothalamic POMC Neurons Mediates Regular Exercise-Induced High-Turnover Metabolism.
Authors:
Journal:Cell Metab
PubMed ID:33535098
Anti-inflammatory effect of strawberry extract against LPS-induced stress in RAW 264.7 macrophages.
Authors:Gasparrini M, Forbes-Hernandez TY, Giampieri F, Afrin S, Alvarez-Suarez JM, Mazzoni L, Mezzetti B, Quiles JL, Battino M
Journal:Food Chem Toxicol
PubMed ID:28130090
'A common denominator in the pathogenesis of most chronic inflammatory diseases is the involvement of oxidative stress, related to ROS production by all aerobic organisms. Dietary antioxidants from plant foods represent an efficient strategy to counteract this condition. The aim of the present study was to evaluate the protective effects ... More
Targeting Breast Cancer Stem Cell State Equilibrium through Modulation of Redox Signaling.
Authors:
Journal:Cell Metab
PubMed ID:29972798
Integrative Omics Analyses Reveal Epigenetic Memory in Diabetic Renal Cells Regulating Genes Associated With Kidney Dysfunction.
Authors:
Journal:Diabetes
PubMed ID:32747424