CellLight™ ER-GFP, BacMam 2.0
Invitrogen™

CellLight™ ER-GFP, BacMam 2.0

CellLight ER-GFP, BacMam 2.0, provides an easy way to label endoplasmic reticulum (ER) with green fluorescent protein (GFP) in liveRead more
Have Questions?
Catalog NumberQuantity
C105901 mL
Catalog number C10590
Price (USD)
570.72
Special offer
Online exclusive
Ends: 31-Dec-2026
696.00
Save 125.28
Each
Quantity:
1 mL
Price (USD)
570.72
Special offer
Online exclusive
Ends: 31-Dec-2026
696.00
Save 125.28
Each
CellLight ER-GFP, BacMam 2.0, provides an easy way to label endoplasmic reticulum (ER) with green fluorescent protein (GFP) in live cells. Simply add the reagent to your cells, incubate overnight, and the cells are ready to image in the morning.

Want to label other cell structures? Learn more about CellLight fluorescent protein labeling tools

This ready-to-use construct is transfected into cells using BacMam 2.0 technology and expresses GFP fused to the ER signal sequence of calreticulin and KDEL (ER retention signal) with minimal cellular disruption. You can observe ER-GFP behavior in live cells using fluorescent imaging and multiplex with other fluorescent proteins or organic dyes.

Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexed imaging using immunocytochemical techniques.

CellLight Technology is:
• Fast and convenient: simply add CellLight reagent to your cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
• Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
• Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
• Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling

BacMam Technology
CellLight ER-GFP, BacMam 2.0, is a fusion construct of ER signal sequence of calreticulin and KDEL (ER retention signal) and emGFP, providing accurate and specific targeting to cellular ER-GFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Detection MethodFluorescence
Dye TypeGFP (EmGFP)
EmissionVisible
Excitation Wavelength Range488⁄510
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope
FormLiquid
Product LineCellLight
Quantity1 mL
Shipping ConditionWet Ice
TechniqueFluorescence Intensity
Label TypeFluorescent Dye
Product TypeEndoplasmic Reticulum Label
Sub Cellular LocalizationEndoplasmic Reticulum
Unit SizeEach
Contents & Storage
Store at 2°C to 6°C, protected from light. Do Not Freeze.

Frequently asked questions (FAQs)

How can I increase the transduction efficiency with the BacMam 2.0 reagents such as the the CellLight and Premo products?

Try varying particle-to-cell ratio (PPC), incubation volume, temperature and, cell density (if adherent cells are transduced). For adherent cells, we recommend a confluence of about 70%. Following the PPC, adjusting the volume is the next best parameter to change to optimize protein expression. If that doesn't work, you can also use the BacMam Enhancer Kit (Cat. No. B10107).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Is there any way to preserve the CellLights labeling beyond 5 days?

Cells transduced with the CellLights reagents can be stored frozen for several months after transduction, without loss of expression.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the CellLights products toxic to cells?

If the viral particles are used at the level we recommend, they are very well tolerated by cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

For how long will the CellLights products label my cells?

The BacMam 2.0 CellLights typically express for 5 days after transduction.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What cell types can the CellLights products be used with?

The first generation BacMam reagents were shown to efficiently transduce over 90 cell types, including stable cell lines and primary cells. With BacMam 2.0, it is now possible to also efficiently transduce primary neurons and stem cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (26)

Citations & References
Abstract
Mutations in ABCD4 cause a new inborn error of vitamin B12 metabolism.
Authors:Coelho D, Kim JC, Miousse IR, Fung S, du Moulin M, Buers I, Suormala T, Burda P, Frapolli M, Stucki M, Nürnberg P, Thiele H, Robenek H, Höhne W, Longo N, Pasquali M, Mengel E, Watkins D, Shoubridge EA, Majewski J, Rosenblatt DS, Fowler B, Rutsch F, Baumgartner MR,
Journal:Nat Genet
PubMed ID:22922874
Inherited disorders of vitamin B12 (cobalamin) have provided important clues to how this vitamin, which is essential for hematological and neurological function, is transported and metabolized. We describe a new disease that results in failure to release vitamin B12 from lysosomes, which mimics the cblF defect caused by LMBRD1 mutations. ... More
Intracellular trafficking of Clostridium perfringens iota-toxin b.
Authors:Nagahama M, Umezaki M, Tashiro R, Oda M, Kobayashi K, Shibutani M, Takagishi T, Ishidoh K, Fukuda M, Sakurai J,
Journal:Infect Immun
PubMed ID:22825447
'Clostridium perfringens iota-toxin is composed of an enzymatic component (Ia) and a binding component (Ib). Ib binds to a cell surface receptor, undergoes oligomerization in lipid rafts, and binds Ia. The resulting complex is then endocytosed. Here, we show the intracellular trafficking of iota-toxin. After the binding of the Ib ... More
Single-cell redox imaging demonstrates a distinctive response of dopaminergic neurons to oxidative insults.
Authors:Horowitz MP, Milanese C, Di Maio R, Hu X, Montero LM, Sanders LH, Tapias V, Sepe S, van Cappellen WA, Burton EA, Greenamyre JT, Mastroberardino PG,
Journal:Antioxid Redox Signal
PubMed ID:21395478
'The study of the intracellular oxido-reductive (redox) state is of extreme relevance to the dopamine (DA) neurons of the substantia nigra pars compacta. These cells possess a distinct physiology intrinsically associated with elevated reactive oxygen species production, and they selectively degenerate in Parkinson''s disease under oxidative stress conditions. To test ... More
Intracellular trafficking mechanism, from intracellular uptake to extracellular efflux, for phospholipid/cholesterol liposomes.
Authors:Un K, Sakai-Kato K, Oshima Y, Kawanishi T, Okuda H,
Journal:Biomaterials
PubMed ID:22858002
'Liposomes are widely used as drug delivery vehicles to transfer chemotherapeutic agents, proteins, and nucleic acids into target cells. To improve therapeutic effects and reduce unexpected toxic side-effects, it is necessary to understand the mechanism of liposomal uptake into cells, and the intracellular fate of internalized liposomes. The intracellular fate ... More
Der p2 Internalization by Epithelium Synergistically Augments Toll-like Receptor-Mediated Proinflammatory Signaling.
Authors:Yin SC, Liao EC, Chiu CL, Chang CY, Tsai JJ,
Journal:
PubMed ID:25749775
'House-dust-mite (HDM) major allergen Der p2 shares homology and function with Toll-like receptor (TLR) signaling protein myeloid differentiation-2 (MD2) and may lead to airway inflammation. Should Der p2 be internalized by human airway epithelium, it has the theoretical propensity to potentiate epithelium activation. This study aimed to demonstrate the internalization ... More