CellLight™ Golgi-RFP, BacMam 2.0
Invitrogen™

CellLight™ Golgi-RFP, BacMam 2.0

CellLight Golgi-RFP, BacMam 2.0, provides an easy way to label golgi with red fluorescent protein (RFP) in live cells. SimplyRead more
Have Questions?
Catalog NumberQuantity
C105931 mL
Catalog number C10593
Price (USD)
577.28
Special offer
Online exclusive
Ends: 31-Dec-2026
704.00
Save 126.72
Each
Quantity:
1 mL
Price (USD)
577.28
Special offer
Online exclusive
Ends: 31-Dec-2026
704.00
Save 126.72
Each
CellLight Golgi-RFP, BacMam 2.0, provides an easy way to label golgi with red fluorescent protein (RFP) in live cells. Simply add the reagent to your cells, incubate overnight, and the cells are ready to image in the morning.

Want to label other cell structures? Learn more about CellLight fluorescent protein labeling tools

This ready-to-use construct is transfected into cells using BacMam 2.0 technology, where it expresses RFP fused to human golgi resident enzyme (N-acetylgalactosaminyltransferase). You can observe golgi-RFP behavior in live cells using fluorescent imaging and multiplex with other fluorescent proteins or organic dyes.

Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexed imaging using immunocytochemical techniques.

CellLight Technology is:
• Fast and convenient: simply add CellLight reagent to your cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
• Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
• Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
• Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling

BacMam Technology
CellLight Golgi-RFP, BacMam 2.0, is a fusion construct of human golgi resident enzyme (N-acetylgalactosaminyltransferase) and TagRFP, providing accurate and specific targeting to cellular golgi-RFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorRed-Orange, Orange
Detection MethodFluorescence
Dye TypeRFP (TagRFP)
EmissionVisible
Excitation Wavelength Range555⁄584
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope
FormLiquid
Product LineCellLight
Quantity1 mL
Shipping ConditionWet Ice
TechniqueFluorescence Intensity
Label TypeFluorescent Dye
Product TypeGolgi Label
Sub Cellular LocalizationGolgi
Unit SizeEach
Contents & Storage
Store at 2°C to 6°C, protected from light. Do Not Freeze.

Frequently asked questions (FAQs)

How can I increase the transduction efficiency with the BacMam 2.0 reagents such as the the CellLight and Premo products?

Try varying particle-to-cell ratio (PPC), incubation volume, temperature and, cell density (if adherent cells are transduced). For adherent cells, we recommend a confluence of about 70%. Following the PPC, adjusting the volume is the next best parameter to change to optimize protein expression. If that doesn't work, you can also use the BacMam Enhancer Kit (Cat. No. B10107).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Is there any way to preserve the CellLights labeling beyond 5 days?

Cells transduced with the CellLights reagents can be stored frozen for several months after transduction, without loss of expression.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the CellLights products toxic to cells?

If the viral particles are used at the level we recommend, they are very well tolerated by cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

For how long will the CellLights products label my cells?

The BacMam 2.0 CellLights typically express for 5 days after transduction.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What cell types can the CellLights products be used with?

The first generation BacMam reagents were shown to efficiently transduce over 90 cell types, including stable cell lines and primary cells. With BacMam 2.0, it is now possible to also efficiently transduce primary neurons and stem cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (31)

Citations & References
Abstract
Faecal excretion of ciprofloxacin after a single oral dose and its effect on faecal bacteria in healthy volunteers.
Authors:Pecquet S, Ravoire S, Andremont A,
Journal:J Antimicrob Chemother
PubMed ID:2211433
'High concentrations of ciprofloxacin have been shown to persist in the faeces of volunteers for several days after a week of oral treatment with this drug, which was also found to have a prolonged effect on aerobic Gram-negative intestinal bacteria. To determine whether a shorter course of ciprofloxacin would have ... More
Intracellular trafficking mechanism, from intracellular uptake to extracellular efflux, for phospholipid/cholesterol liposomes.
Authors:Un K, Sakai-Kato K, Oshima Y, Kawanishi T, Okuda H,
Journal:Biomaterials
PubMed ID:22858002
'Liposomes are widely used as drug delivery vehicles to transfer chemotherapeutic agents, proteins, and nucleic acids into target cells. To improve therapeutic effects and reduce unexpected toxic side-effects, it is necessary to understand the mechanism of liposomal uptake into cells, and the intracellular fate of internalized liposomes. The intracellular fate ... More
Utilization of fluorescent probes for the quantification and identification of subcellular proteomes and biological processes regulated by lipid peroxidation products.
Authors:Cummins TD, Higdon AN, Kramer PA, Chacko BK, Riggs DW, Salabei JK, Dell'italia LJ, Zhang J, Darley-Usmar VM, Hill BG,
Journal:Free Radic Biol Med
PubMed ID:22954622
Oxidative modifications to cellular proteins are critical in mediating redox-sensitive processes such as autophagy, the antioxidant response, and apoptosis. The proteins that become modified by reactive species are often compartmentalized to specific organelles or regions of the cell. Here, we detail protocols for identifying the subcellular protein targets of lipid ... More
Quadriwave lateral shearing interferometry for quantitative phase microscopy of living cells.
Authors:Bon P, Maucort G, Wattellier B, Monneret S,
Journal:Opt Express
PubMed ID:19654713
Phase imaging with a high-resolution wavefront sensor is considered. This is based on a quadriwave lateral shearing interferometer mounted on a non-modified transmission white-light microscope. The measurement technology is explained both in the scope of wave optics and geometrical optics in order to discuss its implementation on a conventional microscope. ... More
Combination of Cl-IB-MECA with paclitaxel is a highly effective cytotoxic therapy causing mTOR-dependent autophagy and mitotic catastrophe on human melanoma cells.
Authors:Soares AS, Costa VM, Diniz C, Fresco P,
Journal:
PubMed ID:24659394
Metastatic melanoma is the deadliest form of skin cancer. It is highly resistant to conventional therapies,particularly to drugs that cause apoptosis as the main anticancer mechanism. Recently, induction of autophagic cell death is emerging as a novel therapeutic target for apoptotic-resistant cancers. We aimed to investigate the underlying mechanisms elicited ... More