CellEvent™ Senescence Green Detection Kit
Invitrogen™

CellEvent™ Senescence Green Detection Kit

The CellEvent Senescence Green Detection Kit consists of a fluorescent probe and optimized buffer that enable the image-based detection ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
C10851100 μL
C1085025 μL
Catalog number C10851
Price (USD)
697.65
Online exclusive
758.00
Save 60.35
Each
Quantity:
100 μL
Price (USD)
697.65
Online exclusive
758.00
Save 60.35
Each
The CellEvent Senescence Green Detection Kit consists of a fluorescent probe and optimized buffer that enable the image-based detection of senescent cells. The fluorescein-based probe contains two galactoside moieties, making it a target for β-galactosidase. Activation of β-galactosidase is commonly used as a biomarker for senescent cells. This hydrolase enzyme resides in lysosomes and converts β-galactosides into monosaccharides under acidic pH conditions. The enzymatically cleaved product is retained within the cell due to covalent binding of intracellular proteins and emits a fluorogenic signal that has absorption/emission maxima of 490/514 nm.

CellEvent Senescence Green Detection Kit features include:
• Reliable and quick fluorescent detection of senescent cells
• Greater sensitivity than traditional colorimetric x-gal
• Fluorescent signal is well retained and detected using standard Alexa Fluor 488/FITC filter sets
• Multiplex-enabled—fluorescent senescence probe can be multiplexed

Due to a limited replicative lifespan, normal cells enter cell cycle arrest, also known as cellular senescence. While in this senescent phase the cells remain metabolically active without undergoing cell death or division. They adopt a specific phenotypic that includes the appearance of multiple nuclei, increased vacuolization, expression of pH-dependent β-galactosidase, and morphological enlargement and extension. Senescence, through a variety of mechanisms, may play a role in tumor suppression, tumor progression, aging, and tissue repair.

Activation of β-galactosidase is commonly used as a biomarker for senescent cells. This hydrolase enzyme resides in lysosomes and converts β-galactosides into monosaccharides under acidic pH conditions. The activity is optimal at lysosomal pH 4, but conventional assays measure at pH 6. It has been shown that normalized β-galactosidase activity is twice as high in senescent cells as in pre-senescent cells regardless of the pH value used for testing.

A colorimetric substrate for β-galactosidase, 5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside (x-gal), has long been used to detect metabolic activity in cells in vitro. However, its use is limited due to inconsistent signal, length of assay, and inability to multiplex.

Therefore, we have developed a sensitive, fluorescent substrate for β-galactosidase that can be used for the detection of senescent cells. The CellEvent Senescence Green probe is a fluorescein-based reagent that contains two galactoside moieties, making it a specific target of β-galactosidase. The enzyme- cleaved product is retained within the cell due to covalent binding of intracellular proteins and emits a fluorogenic signal that has absorption/emission maxima of 490/514 nm.

The CellEvent Senescent Green Kit provides CellEvent Senescent Green probe and an optimized buffer for the detection of senescent cells in fixed samples. This probe can be multiplexed with other fluorescent reagents compatible with paraformaldehyde fixation.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Detection MethodFluorescence
Format96-well plate
Quantity100 μL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
For Use With (Equipment)Fluorescence Microscope, High Content Analysis Instrument
Product LineCellEvent
Product TypeSenescence Green Detection Kit
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C and protect from light.

Frequently asked questions (FAQs)

Which sample types can I use with the CellEvent Senescence Green Detection Kit?

The CellEvent Senescence Green Detection Kit can be used with various sample types, including fixed cells, adherent cells, and tissue sections.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use the CellEvent Senescence Green Detection Kit (Cat. Nos. C10850, C10851) on already fixed samples, rather than immediately after fixation?

The CellEvent Senescence Green Detection Kit (C10850) requires fixation for the stain to enter lysosomes. The beta-galactosidase remains active immediately following fixation with paraformaldehyde, so there is a requirement to stain the cells shortly after fixation.

We have not tested fixation and long-term storage. The beta-galactosidase activity could decrease if you wait too long after fixation to stain. Therefore, we do not recommend using the CellEvent Senescence Green Detection Kit on already fixed samples.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can the CellEvent Senescence Green Probe be multiplexed using antibodies for intracellular labeling?

Yes. After incubating the cells with CellEvent Senescence Green Probe, permeabilize the cells, wash, and proceed with detection.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use my own buffer with the CellEvent Senescence Green Probe?

We do not recommend using your own buffer with the CellEvent Senescence Green Probe. The assay is pH sensitive and we recommend that you use the buffer provided in the kit be used.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use the CellEvent Senescence Green Probe on live cells?

No. It only works on fixed cells; b-galactosidase is still active immediately following fixation with paraformaldehyde.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.