Concanavalin A (Con A) Conjugates
Invitrogen™

Concanavalin A (Con A) Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugateColorExcitation/EmissionProtein Subtype
C11254Alexa Fluor 350Blue346/442 nmConcanavalin A (Con A)
C11252Alexa Fluor 488Green495/519 nmConcanavalin A (Con A)
C21401Alexa Fluor 488Green495/519 nmConcanavalin A, succinylated
C11253Alexa Fluor 594Red590/617 nmConcanavalin A (Con A)
C21402Alexa Fluor 633Deep Red632/647 nmConcanavalin A (Con A)
C21421Alexa Fluor 647Deep Red650/668 nmConcanavalin A (Con A)
C56126Alexa Fluor Plus 405Violet405/450 nmConcanavalin A (Con A)
C56127Alexa Fluor Plus 750Near-infrared750/777 nmConcanavalin A (Con A)
C825Texas RedRed595/615 nmConcanavalin A (Con A)
C860TetramethylrhodamineOrange555/580 nmConcanavalin A (Con A)
Catalog number C11254
Price (USD)
214.84
Special offer
Online exclusive
Ends: 31-Dec-2026
262.00
Save 47.16
Each
Conjugate:
Alexa Fluor 350
Color:
Blue
Excitation/Emission:
346/442 nm
Protein Subtype:
Concanavalin A (Con A)
Price (USD)
214.84
Special offer
Online exclusive
Ends: 31-Dec-2026
262.00
Save 47.16
Each
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC). 

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorBlue
Cross Reactivityα-man, α-glc
Excitation/Emission346/442 nm
Expression SystemNatural
Label TypeAlexa Fluor Dyes
Ligand TypeLectin
Product LineAlexa Fluor
Protein FamilyLectins
Protein SubtypeConcanavalin A (Con A)
Protein TagNone
Purity or Quality GradeImaging Quality
Quantity5 mg
Research CategoryImaging
Shipping ConditionRoom Temperature
SourceCanavlia ensiformis
ConjugateAlexa Fluor 350
For Use With (Application)Immunocytochemistry, Immunohistochemistry, Immunofluorescence
FormLyophilized
RecombinantNative
SpeciesC. ensiformis
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (4)

Citations & References
Abstract
Assessment of fluorochromes for two-photon laser scanning microscopy of biofilms.
Authors:Neu TR, Kuhlicke U, Lawrence JR
Journal:Appl Environ Microbiol
PubMed ID:11823234
A major limitation for the use of two-proton laser scanning microscopy (2P-LSM) in biofilm and other studies is the lack of a thorough understanding of the excitation-emission responses of potential fluorochromes. In order to use 2P-LSM, the utility of various fluorochromes and probes specific for a range of biofilm constituents ... More
The m6A reader YTHDC2 inhibits lung adenocarcinoma tumorigenesis by suppressing SLC7A11-dependent antioxidant function.
Authors:
Journal:Redox Biol
PubMed ID:33232910
Targeting SLC3A2 subunit of system XC- is essential for m6A reader YTHDC2 to be an endogenous ferroptosis inducer in lung adenocarcinoma.
Authors:
Journal:Free Radic Biol Med
PubMed ID:33785413
Exogenous Monounsaturated Fatty Acids Promote a Ferroptosis-Resistant Cell State.
Authors:
Journal:Cell Chem Biol
PubMed ID:30686757