TaqMan™ 2.5X Lyo-Ready qPCR Master Mix, with excipient
Applied Biosystems™

TaqMan™ 2.5X Lyo-Ready qPCR Master Mix, with excipient

Applied Biosystems TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient is designed to be incorporated directly into a lyophilization process. No additional excipients are required.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
C14081B001500 x 25 μL Reactions
C14081SMP150 x 25 μL Reactions
C14081B0025000 x 25 μL Reactions
Catalog number C14081B001
Price (MXN)
-
No. of Reactions:
500 x 25 μL Reactions

Applied Biosystems TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient is designed to be incorporated directly into a lyophilization process. No additional excipients are required.

Primers and probes of your choice are added to the master mix and the mixture is then lyophilized. A recommended lyophilization program is provided to help reduce the amount of time needed to optimize the lyophilization process. All lots of master mix are functionally tested to help ensure lot-to-lot reproducibility. This reagent is manufactured in compliance with ISO 13485 requirements.

Benefits of TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient include:

  • Ready for lyophilization; guidelines available
  • Optimized for multiplexing—enables up to five-target detection per reaction
  • Tolerance of PCR inhibitors that are typically found in biopharmaceutical, molecular diagnostic, and research applications from both purified samples and crude lysates
  • Lot-to-lot consistency

Optimized for lyophilization

TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient has been optimized for consistent performance before and after lyophilization as shown in the figure below. Comprehensive lyophilization guidelines are included with the product to help shorten your path from liquid to lyophilized product. The proprietary excipient formulation helps ensure fast re-hydration of the lyophilized product after liquid sample is added.

Optimized for multiplexing

TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient has been validated for multiplexing up to five targets in one reaction, allowing for additional assays and/or controls to be run simultaneously for increased efficiency. The figure below illustrates this multiplexing capability by comparing performance between singleplex and multiplex reactions.

Tolerance of inhibitors

The unique proprietary formulation allows robust performance even in the presence of substances that can inhibit PCR, resulting in effective template detection from both purified RNA and crude lysate (see figures below).

Lot-to-lot consistency

TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient has been optimized for highly reproducible detection from a wide variety of samples (see figure below). Another figure below shows the consistent Cq results obtained from three unique lots across multiple assays, to help maximize confidence in your results and highlight the exceptional lot-to-lot consistency of this master mix.

For Use as a Raw Material Only.
Specifications
CompatibilityLyophilization
FormatMaster Mix
Hot StartBuilt-in Hot Start
Inhibitor Tolerance (Simple)Medium
Multiplex CapabilityUp to 5-plex
No. of Reactions500 x 25 μL Reactions
Passive Reference DyeNone
PolymeraseAmpliTaq Fast DNA Polymerase
Quantity5 mL
Sample TypeDNA
SensitivityDown to 5 copies
Shipping ConditionDry Ice
Target SpecificitycDNA or gDNA only
ThermostabilityNot Applicable
Volume1 x 5 mL
Concentration2.5X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Microbial Detection
FormLiquid
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
Store at -15°C to -25°C. Protect from light.