TaqMan™ 2.5X Lyo-Ready qPCR Master Mix, with excipient
Applied Biosystems™

TaqMan™ 2.5X Lyo-Ready qPCR Master Mix, with excipient

Applied Biosystems TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient is designed to be incorporated directly into a lyophilization process. No additional excipients are required.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoN.º de reacciones
C14081B0025000 x 25 μL Reactions
C14081SMP150 x 25 μL Reactions
C14081B001500 x 25 μL Reactions
Número de catálogo C14081B002
Precio (USD)
-
N.º de reacciones:
5000 x 25 μL Reactions

Applied Biosystems TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient is designed to be incorporated directly into a lyophilization process. No additional excipients are required.

Primers and probes of your choice are added to the master mix and the mixture is then lyophilized. A recommended lyophilization program is provided to help reduce the amount of time needed to optimize the lyophilization process. All lots of master mix are functionally tested to help ensure lot-to-lot reproducibility. This reagent is manufactured in compliance with ISO 13485 requirements.

Benefits of TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient include:

  • Ready for lyophilization; guidelines available
  • Optimized for multiplexing—enables up to five-target detection per reaction
  • Tolerance of PCR inhibitors that are typically found in biopharmaceutical, molecular diagnostic, and research applications from both purified samples and crude lysates
  • Lot-to-lot consistency

Optimized for lyophilization

TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient has been optimized for consistent performance before and after lyophilization as shown in the figure below. Comprehensive lyophilization guidelines are included with the product to help shorten your path from liquid to lyophilized product. The proprietary excipient formulation helps ensure fast re-hydration of the lyophilized product after liquid sample is added.

Optimized for multiplexing

TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient has been validated for multiplexing up to five targets in one reaction, allowing for additional assays and/or controls to be run simultaneously for increased efficiency. The figure below illustrates this multiplexing capability by comparing performance between singleplex and multiplex reactions.

Tolerance of inhibitors

The unique proprietary formulation allows robust performance even in the presence of substances that can inhibit PCR, resulting in effective template detection from both purified RNA and crude lysate (see figures below).

Lot-to-lot consistency

TaqMan 2.5X Lyo-Ready qPCR Master Mix with excipient has been optimized for highly reproducible detection from a wide variety of samples (see figure below). Another figure below shows the consistent Cq results obtained from three unique lots across multiple assays, to help maximize confidence in your results and highlight the exceptional lot-to-lot consistency of this master mix.

For Use as a Raw Material Only.
Especificaciones
CompatibilidadLyophilization
FormatoMaster Mix
Inicio en calienteBuilt-in Hot Start
Inhibitor Tolerance (Simple)Medium
Capacidad multiplexUp to 5-plex
N.º de reacciones5000 x 25 μL Reactions
Colorante de referencia pasivaNone
PolimerasaAmpliTaq Fast DNA Polymerase
Cantidad50 mL
Tipo de muestraADN
SensibilidadDown to 5 copies
Condiciones de envíoDry Ice
Especificidad de dianacDNA or gDNA only
TermoestabilidadNot Applicable
Volumen1 x 50 mL
Concentración2.5X
Método de detecciónSonda de cebado
Para utilizar con (aplicación)Gene Expression, Microbial Detection
FormularioLíquido
Método de PCRqPCR
Velocidad de reacciónrápida o estándar
Unit SizeEach
Contenido y almacenamiento
Store at -15°C to -25°C. Protect from light.