본 제품은 LMO 제품으로, 고객 분께서 LMO 신고 시스템을 통해 직접 수입 신고를 진행해주셔야 합니다. 자세히보기
Product Image
Invitrogen™

GS115, Pichia pastoris Yeast Strain

Several Pichia pastoris strains are available to allow you to optimize expression and recovery of your protein of interest. Table자세히 알아보기
Have Questions?
카탈로그 번호수량
C181001 Stab
카탈로그 번호 C18100
제품 가격(KRW)
651,000
온라인 행사
Ends: 31-Mar-2026
743,000
할인액 92,000 (12%)
Each
수량:
1 Stab
제품 가격(KRW)
651,000
온라인 행사
Ends: 31-Mar-2026
743,000
할인액 92,000 (12%)
Each
Several Pichia pastoris strains are available to allow you to optimize expression and recovery of your protein of interest. Table 1 provides information to help you choose the appropriate strain.
For Research Use Only. Not for use in diagnostic procedures.
사양
박테리아 또는 효모 균주GS115
세포주Yeast
제품 유형Pichia pastoris Yeast Strain
수량1 Stab
P. pastoris
Unit SizeEach

자주 묻는 질문(FAQ)

When selecting for blasticidin-resistant transformants in the X-33 strain using pPIC6/pPIC6α vectors, why do I get large and small colonies on YPD plates containing 300 µg/ml blasticidin?

Generally, large colonies represent transformants containing pPIC6/pPIC6α integrants, while small colonies represent transformants containing pPIC6/pPIC6α non-integrants. These non-integrants have transduced the pPIC6/pPIC6α plasmid, and therefore, exhibit a low level of blasticidin resistance in the initial selection process. Upon subsequent screening, these non-integrant transformants do not retain blasticidin resistance.

When choosing a blasticidin-resistant transformant for your expression studies, we recommend that you pick blasticidin-resistant colonies from the initial transformation plate and streak them on a second YPD plate containing the appropriate concentration of blasticidin. Select transformants that remain blasticidin-resistant for further studies.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My transformation is not working. Do you have any suggestions?

Here are some suggestinos:

- Make sure that you have harvested cells during log-phase growth (OD <1.0 generally).
- If electroporation is being used, see the electroporator manual for suggested conditions. Vary electroporation parameters if necessary.
- Use more DNA.
- Use freshly made competent cells.
- If the LiCl transformation method is being used, try boiling the carrier DNA.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My spheroplasting of Pichia worked twice, but hasn't worked since. The OD of the culture simply does not drop.

Here are some things to consider:

- If the OD of cells that are used is too high, they will not spheroplast. Do not overgrow cells.
- Do not use old cells and make sure that they are in log phase of growth.
- Make sure to mix zymolyase well before using. Zymolyase is more of a suspension than a solution.
- Make the PEG solution fresh each time and check the pH.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

What are the different kinds of media used for culturing Pichia pastoris and S. cerevisiae?

Following are the rich and minimal media used for culturing Pichia pastoris and S. cerevisiae:

Rich Media:
S. cerevisiae and Pichia pastoris
YPD (YEPD): yeast extract, peptone, and dextrose
YPDS: yeast extract, peptone, dextrose, and sorbitol

Pichia pastoris only
BMGY: buffered glycerol-complex medium
BMMY: buffered methanol-complex medium

Minimal Media (also known as drop-out media):
S. cerevisiae
SC (SD): Synthetic complete (YNB, dextrose (or raffinose or galactose), and amino acids)

Pichia pastoris
MGY: minimal glycerol medium
MD: minimal dextrose
MM: minimal methanol
BMGH: buffered minimal glycerol
BMMH: buffered minimal methanol

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Is there a recommended protocol for fermentation using constitutive expression vectors such as pGAPZ?

Use the following high cell density protocol for pGAP clones. Feed carbon until the desired density is reached (300 to 400 g/L wet cell weight (WCW)). If the protein is well-behaved in the fermenter, increase to 300-400 g/L WCW as with methanol inducible clones. These densities can be reached in less than 48 hours of fermentation. We have fermented constitutive expressers on glycerol using these protocols with good results. Some modifications to the Fermentation Basal Salts Medium that you might want to make are:

1) Substitute 2% dextrose for the 4% glycerol in the batch medium.
2) Substitute 40% dextrose for the 50% glycerol in the fed-batch medium.
3) Feed the 40% dextrose at 12 mL/L/hr (Jim Cregg has published data on expression using several carbon sources as substrates; dextrose gave the highest levels of expression).
4) Yeast extract and peptone may be added to the medium for protein stability.

One warning: If you are working with His- strains, they remain His- after transformation with pGAPZ. Fermentation in minimal medium will require addition of histidine to the fermenter.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

인용 및 참조 문헌 (5)

인용 및 참조 문헌
Abstract
Rhodopsin kinase: expression in mammalian cells and a two-step purification.
Authors:Bruel C, Cha K, Reeves PJ, Getmanova E, Khorana HG
Journal:Proc Natl Acad Sci U S A
PubMed ID:10737781
'A suitable system for expression of the rhodopsin kinase (RK) gene and its mutants is needed for structure-function studies of RK. Previously, investigation of the baculovirus system showed satisfactory production of RK, but posttranslational isoprenylation was deficient. We now report on a comparative study of expression of the RK gene ... More
Biochemical and antigenic characterization of a new dipeptidyl- peptidase isolated from Aspergillus fumigatus.
Authors:Beauvais A, Monod M, Debeaupuis JP, Diaquin M, Kobayashi H, Latge JP
Journal:J Biol Chem
PubMed ID:9045640
A novel dipeptidyl-peptidase (DPP V) was purified from the culture medium of Aspergillus fumigatus. This is the first report of a secreted dipeptidyl-peptidase. The enzyme had a molecular mass of 88 kDa and contained approximately 9 kDa of N-linked carbohydrate. The expression and secretion of dipeptidyl-peptidase varied with the growth ... More
Expression of recombinant HLA-DR2 molecules. Replacement of the hydrophobic transmembrane region by a leucine zipper dimerization motif allows the assembly and secretion of soluble DR alpha beta heterodimers.
Authors:Kalandadze A, Galleno M, Foncerrada L, Strominger JL, Wucherpfennig KW
Journal:J Biol Chem
PubMed ID:8702739
Major histocompatibility complex (MHC) class II molecules are membrane-anchored heterodimers that present peptides on the surface of antigen presenting cells to T cells. Soluble HLA-DR2 molecules were expressed for structural and functional characterization of the MHC/peptide/T cell receptor recognition unit. The alpha and beta chains of DR2 (encoded by the ... More
Potency and selectivity of the cathepsin L propeptide as an inhibitor of cysteine proteases.
Authors:Carmona E, Dufour E, Plouffe C, Takebe S, Mason P, Mort JS, Menard R
Journal:Biochemistry
PubMed ID:8679567
The cathepsin L propeptide (phcl-2) was expressed in Saccharomyces cerevisiae using a human procathepsin L/alpha-factor fusion construct containing a stop codon at position -1 (the C-terminal amino acid of the proregion). Since the yield after purification was very low, the cathepsin L propeptide was also obtained by an alternate procedure ... More
Manipulating monomer-dimer equilibrium of bovine Beta -lactoglobulin by amino acid substitution.
Authors: Sakurai Kazumasa; Goto Yuji;
Journal:J Biol Chem
PubMed ID:12006601
Bovine beta-lactoglobulin, a major protein in cow's milk composed of nine beta-strands (betaA-betaI) and one alpha-helix, exists as a dimer at neutral pH while it dissociates to a native monomer below pH 3.0. It is assumed that the intermolecular beta-sheet formed between I-strands and salt bridges at AB-loops play important ... More