Get the sharpest, brightest images. Try new Invitrogen™ ProLong™ Glass antifade mountant with glass-matching refractive index of 1.52. Learn more
Invitrogen™

CoverWell™ Perfusion Chamber Gasket, 1 chamber, 32 mm x 19 mm, 1.0 mm deep

Catalog number:  C18121

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CoverWell perfusion chamber gaskets are designed to facilitate live-cell imaging and manipulation. These gaskets form watertight Φpress-to-sealΦ chambers with dual-access ports so reagents can be quickly added and removed. Available in single- or multi-well configurations, these gaskets allow multiple experiments to be performed on a single microscope slide or coverslip. The heat-resistant gaskets can be sterilized and used for direct culturing of cells and tissues.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Depth: 1 mm
Dimensions: 32 mm x 19 mm
Product Line: CoverWell™
Product Size: 1 package
Shipping Condition: Room Temperature
Type: Gaskets

Contents & storage

Store at room temperature. 40 per package

Documents

Manuals & protocols

Recommended products