5(6)-CFDA (5-(and-6)-Carboxyfluorescein Diacetate), mixed isomers
Invitrogen™

5(6)-CFDA (5-(and-6)-Carboxyfluorescein Diacetate), mixed isomers

5(6)-CFDA is a cell-permeant esterase substrate that can serve as a viability probe that measures both enzymatic activity, which is자세히 알아보기
Have Questions?
카탈로그 번호수량
C195100 mg
카탈로그 번호 C195
제품 가격(KRW)
337,000
온라인 행사
Ends: 30-Sep-2026
385,000
할인액 48,000 (12%)
Each
수량:
100 mg
제품 가격(KRW)
337,000
온라인 행사
Ends: 30-Sep-2026
385,000
할인액 48,000 (12%)
Each
5(6)-CFDA is a cell-permeant esterase substrate that can serve as a viability probe that measures both enzymatic activity, which is require to activate its fluorescence, and cell-membrane integrity, which is required for intracellular retention of their fluorescent product. Upon hydrolysis by intracellular esterases, this AM ester also yields carboxyfluorescein. As compared with fluorescein, carboxyfluorescein contains extra negative charges and is therefore better retained in cells.
For Research Use Only. Not for use in diagnostic procedures.
사양
염료 유형Other Label(s) or Dye(s)
수량100 mg
시약 유형CFDA & Related Compounds
배송 조건Room Temperature
제품 유형5(6)-CFDA
Unit SizeEach
구성 및 보관
Store in freezer -5°C to -30°C.

인용 및 참조 문헌 (185)

인용 및 참조 문헌
Abstract
Bacteria in post-glacial freshwater sediments.
Authors:Miskin I, Rhodes G, Lawlor K, Saunders JR, Pickup RW
Journal:Microbiology
PubMed ID:9782490
Prokaryote communities in post-glacial profundal freshwater sediments of Windermere, representing 10-12,000 years of deposition, were examined for culturability, viability and community structure. The potential for active geochemical cycles was inferred from the presence of specific groups of bacteria. Direct count procedures revealed 10(12) cells (g dry wt sediment)-1 in the ... More
5-Carboxyfluorescein diacetate as a probe for measuring the growth of keratinocytes.
Authors:Hanthamrongwit M, Reid WH, Courtney JM, Grant MH
Journal:Hum Exp Toxicol
PubMed ID:8086226
There is a requirement for a convenient and reliable method for evaluating the growth rate of human keratinocytes cultured on collagen-based substrates. Therefore, three methods of determining cell growth were first used to quantify the growth rate of the well-characterised L929 mouse fibroblast cell line on tissue culture plastic and ... More
Fluorescent indicators of ion concentrations.
Authors:Tsien RY
Journal:Methods Cell Biol
PubMed ID:2538708
Rapid assessment of physiological status in Escherichia coli using fluorescent probes.
Authors:Porter J, Edwards C, Pickup RW
Journal:J Appl Bacteriol
PubMed ID:7592133
'Rapid and direct viability assessment of Escherichia coli in filtered, sterile lake water was possible using multiparameter flow cytometry. Fluorescent dyes were used as probes for different cellular functions (membrane potential, membrane integrity and intracellular enzyme activity), which were correlated with the ability of the cells to respond to nutrient ... More
Intercellular adhesion mediated by human muscle neural cell adhesion molecule: effects of alternative exon use.
Authors:Pizzey JA, Rowett LH, Barton CH, Dickson G, Walsh FS
Journal:J Cell Biol
PubMed ID:2532218
'Mouse 3T3 fibroblasts were permanently transfected with cDNAs encoding isoforms of the neural cell adhesion molecule (N-CAM) present in human skeletal muscle and brain. Parental and transfected cells were then used in a range of adhesion assays. In the absence of external shear forces, transfection with cDNAs encoding either transmembrane ... More