Thermo Scientific™
Cheminert™ Model C2 Microbore Injectors, N60 stainless, manual actuator, 10 ports
 Related applications:
Industrial Chromatography
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5μL sample loop.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • 1/16 in. fittings
  • 0.010 in. ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator
Documents
Manuals & protocols