Click-iT™ Alexa Fluor™ 555 sDIBO Alkyne, 0.5 mg - FAQs
Click-iT™ Alexa Fluor™ 555 sDIBO Alkyne, 0.5 mg - FAQs
View additional product information for Click-iT™ sDIBO Alkynes for copper-free click chemistry - FAQs (C10407, C20022, C20021, C20020, C20025, C20023)
1 product FAQs found
What concentration and solvent do you recommend for making a stock solution of Click-iT sDIBO Alkynes for copper-free click chemistry?
For Click-iT sDIBO Alkynes for coper-free click chemistry (Cat. Nos. C10407, C20020, C20021, C20022, C20023, C20025), we recommend a 2-5 mM stock solution in DMSO.