Kit de etiquetado de proteínas de maleimida EZ-Link™
Thermo Scientific™

Kit de etiquetado de proteínas de maleimida EZ-Link™

The Thermo Scientific EZ-Link Maleimide Protein Labeling Kit provides a convenient method to label proteins with varying amounts maleimide forMás información
Have Questions?
Número de catálogoCantidad
C200451 Kit
Número de catálogo C20045
Precio (MXN)
-
Cantidad:
1 Kit
The Thermo Scientific EZ-Link Maleimide Protein Labeling Kit provides a convenient method to label proteins with varying amounts maleimide for crosslinking or conjugation with a thiol/sulfhydryl-containing moiety. Sulfo-SMCC, contained in this kit, readily reacts with the primary amines on a protein to form a covalently attached maleimide group that can be further reacted with a reactive thiol or free cystine to create diverse bioconjugates. This kit contains sufficient reagents for five labeling reactions of up to 100 μL sample with a concentration of 1–10 mg/mL.

Features of the EZ-Link Maleimide Protein Labeling Kit include:
• Heterobifunctional—reactive towards amines and thiols/sulfhydryls
• Soluble—sulfo-SMCC reagent readily dissolves in water
• Stable—cyclohexane bridge confers added stability to the maleimide group making sulfo-SMCC the ideal crosslinking agent for maleimide activation of proteins

Sulfo-SMCC is a heterobifunctional crosslinker that contains a sulfonated N-hydroxysuccinimide (NHS) ester and maleimide group that allow covalent conjugation of amine- and thiol/sulfhydryl-containing molecules. NHS esters react with primary amines at pH 7–9 to form amide bonds, while maleimides react with sulfhydryl groups at pH 6.5–7.5 to form stable thioether bonds. The cyclohexane ring in the spacer arm of sulfo-SMCC decreases the rate of hydrolysis of the maleimide group compared to similar reagents that do not contain this ring. This feature enables proteins that have been maleimide labeled with sulfo-SMCC to be lyophilized and stored for later conjugation to a sulfhydryl-containing molecule.

Especificaciones
Reactividad químicaAmina-a-sulfhidrilo
Método de etiquetadoEtiqueta química
Cantidad1 Kit
Fracción reactivaSulfo-NHS, maleimida
Condiciones de envíoHielo húmedo
Línea de productosEZ-Link
Unit SizeEach
Contenido y almacenamiento
Contiene 5 viales de sulfo-SMCC, 5 columnas de eliminación de biotina y colorante Zeba, y un paquete de solución salina tamponada con fosfato BupH™.

Almacene el sulfo-SMCC desecado a –20 °C, y las columnas de eliminación de biotina y colorante Zeba a 2–8 °C.