CyQUANT™ LDH Cytotoxicity Assays
Invitrogen™

CyQUANT™ LDH Cytotoxicity Assays

CyQUANT cytotoxicity assays measure cellular toxicity based on the release of cytosolic enzymes (LDH or G6PD) from damaged cells. These cytotoxicity assays utilize either colorimetric or fluorescence detection and allow continuous monitoring of cell death using microplate readers.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityDetection Method
C20300200 AssaysAbsorbance
C20302200 AssaysFluorescence
C203011000 AssaysAbsorbance
C203031000 AssaysFluorescence
Catalog number C20300
Price (USD)
194.65
Online exclusive
201.00
Save 6.35
Each
Quantity:
200 Assays
Detection Method:
Absorbance
Price (USD)
194.65
Online exclusive
201.00
Save 6.35
Each

CyQUANT cytotoxicity assays provide a simple and reliable method to assess chemical or cell-mediated cellular toxicity. The CyQUANT LDH Cytotoxicity Assays and the CyQUANT Cytotoxicity Assay (G6PD Release Assay) use colorimetric- or fluorescence-based detection to measure the release of cytosolic enzymes (LDH or G6PD) from damaged cells. These cytotoxicity assays are optimized for use on microplate readers and can be used for continuous monitoring of cell health over time since cell lysis is not required, making them versatile tools for high-throughput screening and drug discovery applications.

CyQUANT LDH Cytotoxicity Assays are used to measure cytotoxicity through the release of lactate dehydrogenase (LDH), a proven indicator of cellular toxicity. LDH is a cytosolic enzyme found in many cells that is released into the surrounding cell culture medium when the plasma membrane is damaged due to cellular injury, tissue damage, or disease. CyQUANT LDH Assays enable accurate and quantitative measurement of this extracellular LDH release using either colorimetric or fluorescence detection on microplate readers. These LDH assay kits can be used with different cell types, including 3D cell models. Since LDH released into the medium is the indicator of cytotoxicity, these assays can monitor cellular toxicity from the same sample over time.

In the colorimetric CyQUANT LDH Assay (Cat. Nos. C20300 and C20301), extracellular LDH release is quantified by a coupled enzymatic reaction in which LDH catalyzes the conversion of lactate to pyruvate via NAD+ reduction to NADH. Diaphorase then uses NADH to reduce a tetrazolium salt (INT) to a red formazan product that can be measured at 490 nm.

In the fluorescence CyQUANT LDH Assay (Cat. Nos. C20302 and C20303), extracellular LDH release is quantified by a coupled enzymatic reaction in which LDH catalyzes the conversion of lactate to pyruvate via NAD+ reduction to NADH. Diaphorase then uses NADH to reduce resazurin to resorufin that can be detected using an ex/em of 560/590 nm using a fluorescence microplate reader.

Features of the CyQUANT LDH Cytotoxicity Assay

  • Convenient—add-mix-read format for adherent and suspension cells, including 3D cell models
  • Accurate—provides a quantitative measurement of LDH release
  • Flexible—ideal for high-throughput screening, monitor cytotoxicity from the same sample over time
  • Robust—uses stable LDH enzyme activity as a cytotoxic marker
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-impermeant
DescriptionCyQUANT™ LDH Cytotoxicity Assay
Detection MethodAbsorbance
Dye TypeFormazan
Emission Wavelength490 nm
Format96-well plate
Quantity200 Assays
Shipping ConditionDry Ice
Absorbance Wavelength490 nm
ColorRed
For Use With (Application)Cytotoxicity Assay
For Use With (Equipment)Microplate Reader
Product LineCyQUANT
Product TypeLDH Cytotoxicity Assay
Reference Wavelength680 nm
Unit SizeEach
Contents & Storage
• Substrate Mixture, 1 vial lyophilizate
• Assay Buffer, 1 x 600 μL
• Lysis Buffer, 2.5 mL
• Stop Solution, 12 mL
• LDH Positive Control, 6 μL

Store at -5 to -30°C, protect from light

Frequently asked questions (FAQs)

How many assays can I perform using the CyQUANT LDH Cytotoxicity Assay Kit (Cat. No. C20300)?

The CyQUANT LDH Cytotoxicity Assay Kit (Cat. No. C20300) contains sufficient reagents to perform 200 assays in 96-well plates.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the concentration of the LDH Positive Control included in the CyQUANT LDH Cytotoxicity Assay Kit (Cat. Nos. C20300, C20301)?

The concentration of the LDH Positive Control included in the CyQUANT LDH Cytotoxicity Assay Kit (Cat. Nos. C20300, C20301) is proprietary.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I observed a slight color change when I prepared the Reaction Mixture from the CyQUANT LDH Cytotoxicity Assay Kit (Cat. Nos. C20300, C20301) prior to testing my samples. Is this normal?

The CYQUANT LDH Cytotoxicity Assay Kit (Cat. Nos. C20300, C20301) Assay Buffer used to prepare the Reaction Mixture, contains the reaction dye and therefore has some inherent coloration.

The color change should be considerably noticeable with the Positive Control in comparison to the Assay Buffer alone.

Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center.

Can I collect the media from the CyQUANT LDH Cytotoxicity Assay at different time points during the day and then do the assay later in the day?

We do not recommend doing this. The LDH in the media should be tested as soon as possible to avoid the loss of LDH activity. The reaction mixture is stable for 3 to 4 weeks, stored frozen.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I refrigerate or freeze the collected media from the CyQUANT LDH Cytotoxicity Assay and analyze at a later time?

We do not recommend doing this. Refrigeration or freezing and storage over time would cause the loss of LDH activity.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
Cell Painting for cytotoxicity and mode-of-action analysis in primary human hepatocytes
Authors:Ewald J, Titterton K, Bäuerle A. et al.
Journal:Cell Systems
PubMed ID:
Scalable, human-relevant approaches for detecting compound modes of action are needed to improve chemical safety evaluation. Here, we apply image-based profiling (Cell Painting) alongside two cytotoxicity assays in primary human hepatocytes exposed to eight concentrations of 1,085 compounds spanning pharmaceuticals, pesticides, and industrial chemicals. We compared three computational approaches (CellProfiler, ... More