CellTracker™ Fluorescent Probes
Invitrogen™

CellTracker™ Fluorescent Probes

CellTracker™ is a fluorescent dye well suited for monitoring cell movement or location. After loading into cells, the dye is well retained, allowing for multigenerational tracking of cellular movements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityDye Type
C21105 mgCellTracker™ Blue CMAC
C21115 mgCellTracker™ Blue CMHC
C128815 mgCellTracker Blue CMF2HC
C3456520 x 15 μgCellTracker™ Deep Red
C702520 x 50 μgCellTracker Green CMFDA
C29251 mgCellTracker Green CMFDA
C21025 mgCellTracker Green BODIPY
C3455120 x 50 μgCellTracker Orange CMRA
C29271 mgCellTracker Orange CMTMR
C3455220 x 50 μgCellTracker™ Red CMTPX
C100945 x 0.1 mgCellTracker Violet BMQC
Catalog number C2110
Price (USD)
323.90
Special offer
Online exclusive
Ends: 31-Dec-2026
395.00
Save 71.10
Each
Quantity:
5 mg
Dye Type:
CellTracker™ Blue CMAC
Price (USD)
323.90
Special offer
Online exclusive
Ends: 31-Dec-2026
395.00
Save 71.10
Each
Cell movement and location studies require specialized probes that are nontoxic to living cells and well retained, allowing for multigenerational tracking. The CellTracker fluorescent probes are available in a range of fluorescent colors to match instrument lasers and filters, and to accommodate co-staining with antibodies or other cell analysis probes. These dyes are excellent tools for monitoring cell movement, location, proliferation, migration, chemotaxis, and invasion.

Features of the CellTracker dyes include:

  • Easy to use—remove culture media, add dye, incubate 15-45 minutes, and image cells
  • Excellent retention—fluorescent signal retention of >72 hours (typically three to six generations)
  • Ideal tracking dyes—monitor cell movement, location, proliferation, migration, chemotaxis, and invasion
  • Low cytotoxicity—does not affect viability or proliferation
  • Cell Versatility– works with cells in suspension and adherent cells

The CellTracker fluorescent probes have been designed to freely pass through cell membranes; however, once inside the cell are transformed into cell-impermeant reaction products. The CellTracker fluorescence probes (except for CellTracker Deep Red) contain a chloromethyl or bromomethyl group that reacts with thiol groups, utilizing a glutathione S-transferase–mediated reaction. In most cells, glutathione levels are high (up to 10 mM) and glutathione transferase is ubiquitous. CellTracker Deep Red probe contains a succinimidyl ester reactive group, which reacts with amine groups present on proteins.

After conversion to impermeant versions, the CellTracker fluorescent probes are well retained in living cells through several generations. The probes are transferred to daughter cells, but are not transferred to adjacent cells in a population. Cells loaded with the CellTracker fluorescent probes display fluorescence for at least 72 hours and exhibit ideal tracking dye properties—they are stable, nontoxic at working concentrations, well retained in cells, and brightly fluorescent at physiological pH. Additionally, several CellTracker fluorescent probes with various excitation and emission spectra are available allowing for multiplexing in fluorescence imaging. The CellTracker dyes are retained with fixation and permeabilization, enabling their use with antibodies for fluorescence immunostaining applications.

Spectral characteristics of the fluorescent CellTracker probes:

  • CellTracker Blue CMAC—353/466 nm
  • CellTracker Blue CMF2HC—371/464 nm
  • CellTracker Blue CMHC—372/470 nm
  • CellTracker Violet BMQC—415/516 nm
  • CellTracker Green CMFDA—492/517 nm
  • CellTracker Green BODIPY™—522/529 nm
  • CellTracker Orange CMTMR—541/565 nm
  • CellTracker Orange CMR—548/576 nm
  • CellTracker Red CMTPX—577/602 nm
  • CellTracker Deep Red—630/660 nm

Fluorescent CellTracker reagents include the blue-fluorescent chloromethyl derivatives of amino-, hydroxy-, and difluorohydroxycoumarin (CMAC, CMHC and CMF2HC), the green-fluorescent chloromethyl derivatives of fluorescein diacetate (CMFDA) and a BODIPY™ dye, the orange-fluorescent CMTMR and CMRA, and the red-fluorescent CMTPX. CellTracker Blue CMAC, CMHC, and CMF2HC, CellTracker Violet, the violet-fluorescent bromomethyl derivative of coumarin (BMQC), CellTracker Green BODIPY™, CellTracker Orange CMTMR, and CellTracker Red CMTPX do not require enzymatic cleavage to activate their fluorescence, whereas the green CMFDA and orange CMRA do require enzymatic cleavage. The impermeable reaction products of the chloromethyl or bromomethyl coumarins have excellent retention, strong fluorescence, and relatively uniform cytoplasmic staining, making these derivatives potentially useful for correcting motion artifacts in imaging. CMFDA is colorless and non-fluorescent until cytosolic esterases cleave off the acetates, releasing a brightly fluorescent product.

In addition to bright fluorescence and excellent retention, the CellTracker fluorescent probes do not contribute to cytotoxicity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorBlue
DescriptionCellTracker™ Blue CMAC Dye, 5 mg
Dye TypeCellTracker™ Blue CMAC
Emission353 nm
Excitation Wavelength Range466 nm
FormDry Powder
Molecular Weight (g/mol)209.63
Product LineCellTracker
Quantity5 mg
Reagent TypeCell Tracker Compounds, Cell Labeling Reagents
Shipping ConditionRoom Temperature
Label TypeOther Labels or Dyes
Product TypeDye
Sub Cellular LocalizationCytoplasm
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C) and protect from light.

Frequently asked questions (FAQs)

I want to do a cell migration study for around 4 hours and need to fluorescently label the cells with a dye. What do you recommend?

Calcein, AM and FDA (fluorescein diaceate) are examples of some dyes used for this application. Since these dyes are not incorporated or covalently attached to any cellular components, they may have a short retention time as some cell types may actively efflux the dye out of the cells. The CellTracker and CellTrace dyes include either a mild thiol-reactive chloromethyl group or amine-reactive succinnimidyl ester group to allow for covalent binding to cellular components, providing for better retention. As with any reagent, one should empirically determine retention times for the cell type used.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I need a total cell stain, similar to HCS CellMask Blue stain, to label cytoplasm and nuclei in live cells. What do you recommend?

You can use a combination of two dyes with overlapping blue emission. For cytoplasm, you can label the cell with CellTracker Blue CMAC. This can be combined with Hoechst 33342 for nuclei. Both dyes are imaged using a standard DAPI filter set.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I labeled my cells with Calcein, AM, but when I imaged the next day, there was no fluorescence from Calcein. Why?

Calcein, AM is a good choice for cell tracking and as a general cytoplasmic stain. However, it doesn't bind to anything and may be actively pumped out of the cells within a couple hours, which is likely what happened. The retention of Calcein within live cells is dependent upon the inherent properties of the cell type and culture conditions.

For long-term imaging, you may wish to consider a reactive cytoplasmic stains such as CFDA, SE or the CellTracker and CellTrace dyes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can the CellTracker dyes be fixed?

Yes, the CellTracker dyes react with any accessible thiol part of the protein and can be fixed. However, some CellTracker dyes may be attached to small metabolites that can leak from the cell following permeabilization. This can result in decreased fluorescence.

Find additional tips, troubleshooting help, and resources within our Cell Tracing and Tracking Support Center.

I stained two populations of cells, one with CellTracker Green and the other with CellTracker Red, but it looks like there may be crossover of the red dye to the green cells. What is going on?

One possibility is that there is spectral bleedthrough between the dyes. Be sure to check the single-color samples by imaging the red cells in green and imaging the green cells in red, using the optimal imaging settings for the other color. If you see bleedthrough with these controls, then you will have to reduce the dye label concentration to reduce the brightness of the dyes, or choose dyes that are farther apart spectrally. If the issue isn’t bleedthrough, another possibility is that the cells were not adequately washed after staining, allowing some unincorporated dye to remain and label the other cells after they were introduced. Extending washes and wash times should help with this.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (43)

Citations & References
Abstract
Profiling antibody responses by multiparametric analysis of primary B cells.
Authors:Story CM, Papa E, Hu CC, Ronan JL, Herlihy K, Ploegh HL, Love JC,
Journal:Proc Natl Acad Sci U S A
PubMed ID:19004776
'Determining the efficacy of a vaccine generally relies on measuring neutralizing antibodies in sera. This measure cannot elucidate the mechanisms responsible for the development of immunological memory at the cellular level, however. Quantitative profiles that detail the cellular origin, extent, and diversity of the humoral (antibody-based) immune response would improve ... More
Intercellular spreading of Porphyromonas gingivalis infection in primary gingival epithelial cells.
Authors:Yilmaz O, Verbeke P, Lamont RJ, Ojcius DM
Journal:Infect Immun
PubMed ID:16369027
'Porphyromonas gingivalis, an important periodontal pathogen, is an effective colonizer of oral tissues. The organism successfully invades, multiplies in, and survives for extended periods in primary gingival epithelial cells (GECs). It is unknown whether P. gingivalis resides in the cytoplasm of infected cells throughout the infection or can spread to ... More
T cell receptor engagement leads to the recruitment of IBP, a novel guanine nucleotide exchange factor, to the immunological synapse.
Authors:Gupta S, Fanzo JC, Hu C, Cox D, Jang SY, Lee AE, Greenberg S, Pernis AB
Journal:J Biol Chem
PubMed ID:12923183
'Reorganization of the actin cytoskeleton is crucial to the formation and function of the immunological synapse. Rho GTPases are critical mediators of cytoskeletal reorganization, and their activity at the synapse is likely to be stringently regulated. Guanine nucleotide exchange factors (GEFs) belonging to the Dbl family of proteins represent one ... More
Automated image analysis of live/dead staining of the fungus Aureobasidium pullulans on microscope slides and leaf surfaces.
Authors:Nelson CD, Spear RN, Andrews JH
Journal:Biotechniques
PubMed ID:11056819
'An image analysis program and protocol for the identification and enumeration of live versus dead cells of the yeast-like fungus Aureobasidium pullulans was developed for both populations on microscope slides and leaf surfaces. Live cells took up CellTracker Blue, while nonviable cells stained with DEAD Red. Image analysis macro programs ... More
CX3CR1-fractalkine expression regulates cellular mechanisms involved in adhesion, migration, and survival of human prostate cancer cells.
Authors:Shulby SA, Dolloff NG, Stearns ME, Meucci O, Fatatis A
Journal:Cancer Res
PubMed ID:15256432
'Chemokines and their receptors might be involved in the selection of specific organs by metastatic cancer cells. For instance, the CXCR4-SDF-1alpha pair regulates adhesion and migration of breast as well as prostate cancer cells to metastatic sites. In this study, we present the first evidence for the expression of CX3CR1--the ... More