Invitrogen™

CaptAvidin™ Biotin-Binding Protein

Catalog number:  C21385

Related applications:

Protein Assays and Analysis

| Protein Purification & Isolation

Back to top

Description

CaptAvidin™ biotin-binding protein contains a nitrated tyrosine in its biotin-binding site. This chemical modification permits dissociation of the avidin-biotin complex under milder conditions than the traditional avidin-biotin complex. At pH 4.0, CaptAvidin™ biotin-binding protein binds biotin tightly with a Ka of ~109M-1. This association can then be reversed at pH 10, allowing complete dissociation of the avidin-biotin complex. This is in contrast to the harsh conditions (8 M guanidine hydrochloride, pH 1.5) needed to dissociate the avidin-biotin complex.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Shipping Condition: Room Temperature
Label or Dye: Unconjugated

Contents & storage

Store in freezer (-5 to -30°C).