Co-factors
Co-factors
Thermo Scientific™

Co-factors

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
製品番号(カタログ番号)数量製品タイプ濃度
C24341250 μLPolymerase Co-factor100 mM
F-5101250 μLPolymerase Co-factor50 mM
B13880C800 μLMethyl Group Donor5 mM
製品番号(カタログ番号) C24341
価格(JPY)
2,200
キャンペーン価格
Ends: 27-Mar-2026
3,700
割引額 1,500 (41%)
Each
お問い合わせください ›
数量:
250 μL
製品タイプ:
Polymerase Co-factor
濃度:
100 mM

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
研究用途にのみご使用ください。診断目的には使用できません。
仕様
化学物質名または材質Manganese Choride
分子式MnCl2
反応数100 Reactions
製品タイプPolymerase Co-factor
数量250 μL
濃度100 mM
形状Liquid
フォーマットVial
Unit SizeEach