Cholera Toxin Subunit B (Recombinant) Conjugate
Invitrogen™

Cholera Toxin Subunit B (Recombinant) Conjugate

Recombinant cholera toxin B subunit (CT-B) conjugates bind with high affinity to GM1 ganglioside and support neuronal tracing, GM1 labeling, membrane trafficking, and membrane organization studies. Choose from Alexa Fluor™ 488, 555, 594, and 647 fluorescent conjugates, Biotin-XX, or horseradish peroxidase (HRP) to match your detection workflow.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugateQuantity
C34778Alexa Fluor 647100 μg
C22841Alexa Fluor 488500 μg
C34776Alexa Fluor 555100 μg
C22843Alexa Fluor 555500 μg
C34777Alexa Fluor 594100 μg
C22842Alexa Fluor 594500 μg
C34779Biotin100 μg
C34780
also known as C-34780
HRP (Horseradish Peroxidase)100 μg
C34775Alexa Fluor 488100 μg
Catalog number C34778
Price (USD)
287.82
Special offer
Online exclusive
Ends: 31-Dec-2026
351.00
Save 63.18
Each
Conjugate:
Alexa Fluor 647
Quantity:
100 μg
Price (USD)
287.82
Special offer
Online exclusive
Ends: 31-Dec-2026
351.00
Save 63.18
Each

Recombinant cholera toxin B conjugates for GM1 labeling and neuronal tracing

Cholera toxin B subunit (CT-B) binds with high affinity to GM1 ganglioside on the cell surface and is widely used for neuronal tracing, GM1 labeling, endocytosis, and membrane trafficking studies. At neutral pH, CT-B forms a pentamer with multiple GM1-binding sites, enabling multivalent interactions with GM1-containing membranes.

These conjugates are made using recombinant cholera toxin B subunit without the enzymatically active A subunit of native cholera toxin.

Why use recombinant cholera toxin B conjugates?

  • Label GM1 ganglioside: Use the high-affinity interaction between CT-B and GM1 to visualize and detect GM1-associated membrane structures.
  • Trace neuronal projections: Use CT-B for well-established retrograde labeling of neuronal pathways.
  • Study membrane trafficking: Follow CT-B/GM1 internalization and retrograde trafficking through endosomal pathways.
  • Investigate membrane organization: Label GM1 and examine GM1-associated membrane domains in lipid raft-related studies.
  • Use recombinant B subunit: Study GM1 binding and trafficking with CT-B lacking the enzymatically active A subunit of native cholera toxin.
  • Choose multiple detection formats: Select fluorescent Alexa Fluor™ conjugates, Biotin-XX, or HRP to match fluorescence, affinity-based, or enzyme-based detection workflows.

Choose a CT-B conjugate for your detection workflow

Select from Alexa Fluor™ 488, 555, 594, and 647 CT-B conjugates for fluorescence imaging, Biotin-XX for avidin- or streptavidin-based detection, or HRP for enzyme-based detection.

Why choose from multiple conjugates?

  • Match your imaging system: Select an Alexa Fluor™ conjugate compatible with available excitation sources, emission filters, and detectors.
  • Support multicolor imaging: Choose spectrally distinct Alexa Fluor™ dyes for use with other fluorescent probes, antibodies, and cellular markers.
  • Enable affinity-based detection: Use Biotin-XX conjugates with avidin- or streptavidin-based detection workflows.
  • Enable enzyme-based detection: Use HRP conjugates for chromogenic or chemiluminescent detection workflows.
  • Use one GM1-binding probe across workflows: Apply recombinant CT-B with different labels while maintaining the same GM1-binding biological probe.

Applications of cholera toxin B conjugates

Cholera toxin B conjugates are versatile probes for neuronal tracing, GM1 ganglioside labeling, endocytosis, and membrane trafficking studies.

Use CT-B conjugates to:

  • Perform neuronal tracing, including widely used retrograde labeling of neuronal projections
  • Label GM1 ganglioside on cell membranes
  • Visualize GM1-associated membrane domains in lipid raft-related studies
  • Study CT-B/GM1 internalization and retrograde trafficking through endosomal pathways
  • Perform fluorescence imaging with Alexa Fluor™ 488, 555, 594, or 647 conjugates
  • Perform avidin- or streptavidin-based detection with Biotin-XX conjugates
  • Perform enzyme-based detection with HRP conjugates

For Research Use Only. Not for use in diagnostic procedures.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein FormRecombinant
Protein SubtypeCholera Toxin
Quantity100 μg
Shipping ConditionRoom Temperature
ConjugateAlexa Fluor 647
FormLyophilized
RecombinantRecombinant
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

I injected a fluorescent tracer, but cannot detect it after tissue is fixed and sectioned. What am I doing wrong?

Confirm that the tracer you are using crosslinks to proteins or has a primary amine for fixation-either a hydrazide, lysine fixable dextran, or a protein conjugate.
Use aldehyde-based fixatives to cross link the amines on the tracer.
Inject a larger amount or higher concentration of the tracer. Tracers are generally injected at 1-20% concentrations (10 mg/mL or higher).
Confirm that you are using the correct fluorescent filter for detection. You can perform a spot test by pipetting a small amount of the undiluted stock solution of the tracer onto a slide, then view under the filter you are using on your microscope. This will confirm if the tracer fluorescence can be detected and the fluorescent microscope filter is working properly.
Review tissue fixation and handling procedures to confirm if any reagents or processing procedures could be affecting the tracer.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Do you have a tracer that will only transport retrograde?

Wheat germ agglutinin and cholera toxin conjugates have been used for retrograde tracing. They may have some anterograde tracing in some applications. A selection guide can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing/protein-conjugates.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How do I know which tracer to choose for my experiment?

Factors to consider are size of tracer, method of delivery (injection, direct application to tissue, etc.), and if the tracer needs to be fixable. Here are some links to details about the various classes of neuronal tracers we offer and how to choose between them:

Neuronal Tracing (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html)
Choosing a Tracer (https://www.thermofisher.com/us/en/home/references/molecular-probes-the-handbook/fluorescent-tracers-of-cell-morphology-and-fluid-flow/choosing-a-tracer.html)
Imaging Analysis (http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/bioprobes-50-journal.pdf)

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What products do you have for neuronal tracing?

Please check out this web page (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html) for details.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (27)

Citations & References
Abstract
Inhibition of caveolar uptake, SV40 infection, and beta1-integrin signaling by a nonnatural glycosphingolipid stereoisomer.
Authors:Singh RD, Holicky EL, Cheng ZJ, Kim SY, Wheatley CL, Marks DL, Bittman R, Pagano RE
Journal:J Cell Biol
PubMed ID:17371832
'Caveolar endocytosis is an important mechanism for the uptake of certain pathogens and toxins and also plays a role in the internalization of some plasma membrane (PM) lipids and proteins. However, the regulation of caveolar endocytosis is not well understood. We previously demonstrated that caveolar endocytosis and beta1-integrin signaling are ... More
Actin depolymerization transduces the strength of B-cell receptor stimulation.
Authors:Hao S, August A
Journal:Mol Biol Cell
PubMed ID:15728723
'Polymerization of the actin cytoskeleton has been found to be essential for B-cell activation. We show here, however, that stimulation of BCR induces a rapid global actin depolymerization in a BCR signal strength-dependent manner, followed by polarized actin repolymerization. Depolymerization of actin enhances and blocking actin depolymerization inhibits BCR signaling, ... More
Silica crystals and aluminum salts activate the NALP3 inflammasome through phagosomal destabilization.
Authors:Hornung V, Bauernfeind F, Halle A, Samstad EO, Kono H, Rock KL, Fitzgerald KA, Latz E,
Journal:Nat Immunol
PubMed ID:18604214
'Inhalation of silica crystals causes inflammation in the alveolar space. Prolonged exposure to silica can lead to the development of silicosis, an irreversible, fibrotic pulmonary disease. The mechanisms by which silica and other crystals activate immune cells are not well understood. Here we demonstrate that silica and aluminum salt crystals ... More
Bacterial RNA:DNA hybrids are activators of the NLRP3 inflammasome.
Authors:Kailasan Vanaja S, Rathinam VA, Atianand MK, Kalantari P, Skehan B, Fitzgerald KA, Leong JM,
Journal:
PubMed ID:24828532
'Enterohemorrhagic Escherichia coli (EHEC) is an extracellular pathogen that causes hemorrhagic colitis and hemolytic uremic syndrome. The proinflammatory cytokine, interleukin-1ß, has been linked to hemolytic uremic syndrome. Here we identify the nucleotide-binding domain and leucine rich repeat containing family, pyrin domain containing 3 (NLRP3) inflammasome as an essential mediator of ... More
Tracing from fat tissue, liver, and pancreas: a neuroanatomical framework for the role of the brain in type 2 diabetes.
Authors:Kreier F, Kap YS, Mettenleiter TC, van Heijningen C, van der Vliet J, Kalsbeek A, Sauerwein HP, Fliers E, Romijn JA, Buijs RM
Journal:Endocrinology
PubMed ID:16339209
'The hypothalamus uses hormones and the autonomic nervous system to balance energy fluxes in the body. Here we show that the autonomic nervous system has a distinct organization in different body compartments. The same neurons control intraabdominal organs (intraabdominal fat, liver, and pancreas), whereas sc adipose tissue located outside the ... More