CellTrace™ Calcein Red-Orange, AM - Special Packaging
Invitrogen™
CellTrace™ Calcein Red-Orange, AM - Special Packaging
Calceína AM de color rojo-anaranjado CellTrace™, AM; embalaje especial
Have Questions?
Número de catálogoCantidad
C3485120 x 50 μg
Número de catálogo C34851
Precio (MXN)
-
Cantidad:
20 x 50 μg
CellTrace calcein red-orange AM is a cell-permeant dye that can be used to determine cell viability in most eukaryotic cells. Unlike calcein AM (C-1430, C-3099, C-3100), CellTrace calcein red-orange AM is intrinsically fluorescent; thus, an additional wash step may be necessary to minimize background fluorescence from dye that is not taken up by cells. However, CellTrace calcein red-orange (excitation/emission maxima 577/590 nm) is well-retained by live cells that possess intact plasma membranes, and consequently it is a useful cell tracer and indicator of cell viability.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Permeabilidad celularPermeabilidad celular
DescripciónCalceína AM de color rojo-anaranjado, AM; embalaje especial
Tipo de coloranteOtras etiquetas o colorantes
Longitud de onda de emisión590 nm
Longitud de onda de excitación577 nm
FormularioLiofilizado
Cantidad20 x 50 μg
Tipo de reactivoCompuestos de seguimiento celular, reactivos de etiquetado celular
Condiciones de envíoTemperatura ambiente
Enzima dianaEsterasa
Para utilizar con (aplicación)Rastreo celular, Rastreador celular
Para utilizar con (equipo)Microscopio de fluorescencia
Línea de productosCellTrace
Tipo de productoTinte
Unit SizeEach
Contenido y almacenamiento
Almacenar en el congelador de -5 °C a -30 °C y proteger de la luz.
Preguntas frecuentes
I need a general cytoplasmic stain that does not overlap with the GFP in my cells. What do you recommend?
Calcein AM, a green dye, is typically used as a general cytoplasmic stain, but not recommended with GFP-positive cells. For GFP-expressing cells there are other colors available: Calcein Blue AM, Calcein Violet AM, and Calcein Red-Orange AM. The retention time of these dyes in live cells is dependent upon the inherent properties of the cell.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
How do I reconstitute CellTrace Calcein Red-Orange, AM (Cat. No. C34851)?
You can dissolve the dye using high-quality, anhydrous dimethylsulfoxide (DMSO) up to 1 -2 mg/mL. The acetoxymethyl ester (AM) moiety on CellTrace Calcein Red-Orange, AM is susceptible to hydrolysis when exposed to water absorbed by the DMSO. Once prepared, use the DMSO stock solutions of CellTrace Calcein Red-Orange, AM within a short time period. Aqueous working solutions containing the dye should be prepared fresh and used on the same day. The working principle of CellTrace Calcein Red-Orange, AM is very similar to Calcein, AM, cell-permeant dye (Cat. No. C1430). The protocol given in the product manual for Calcein, AM with an additional wash step is suitable. Wash cells with pre-warmed buffer (e.g. PBS, HBSS) to remove residual serum present in the culture medium, load cells with reagent in buffer, incubate from 15-45 minutes and then wash cells with medium (with or without serum). For further information please see the User Guide.
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center.
I stained two populations of cells, one with CellTracker Green and the other with CellTracker Red, but it looks like there may be crossover of the red dye to the green cells. What is going on?
One possibility is that there is spectral bleedthrough between the dyes. Be sure to check the single-color samples by imaging the red cells in green and imaging the green cells in red, using the optimal imaging settings for the other color. If you see bleedthrough with these controls, then you will have to reduce the dye label concentration to reduce the brightness of the dyes, or choose dyes that are farther apart spectrally. If the issue isn’t bleedthrough, another possibility is that the cells were not adequately washed after staining, allowing some unincorporated dye to remain and label the other cells after they were introduced. Extending washes and wash times should help with this.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I stained my cells with Calcein, AM, but the signal went away after I fixed my cells. Why is this?
Calcein, AM diffuses into cells, the 'AM' moiety is cleaved by cellular esterases, and then the dye molecules are observed in the cytoplasm without binding to anything. This gives a 'whole cell' stain. It also means that the dyes are not crosslinked with aldehyde-based fixation and thus will be lost upon fixation. Additionally, any disruption of plasma membrane, such as with detergents or trypsinization, will lead to leakage of the dye from the cell.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I'm trying to stain my cells with CellTracker dyes or CFDA SE, but I'm not seeing much signal. What can I do?
First, make sure you aren’t staining in the presence of serum, since serum can have esterase activity that can prematurely cleave the AM group on these dyes, preventing entry into cells. After staining, it’s okay to return the cells to medium containing serum. After this, you can try increasing the concentration and label time to get a higher intensity.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Citations & References (11)
Citations & References
Abstract
T lymphocytes expressing a CD16 signaling receptor exert antibody-dependent cancer cell killing.
Authors:Kudo K, Imai C, Lorenzini P, Kamiya T, Kono K, Davidoff AM, Chng WJ, Campana D,
Journal:
PubMed ID:24197131
To expand applications for T-cell-based immunotherapy in cancer, we designed a receptor that binds the Fc portion of human immunoglobulins and delivers activation signals. The construct included the high-affinity CD16 (FCGR3A) V158 variant, CD8a hinge, and transmembrane domains, along with signaling domains from CD3? and 4-1BB (TNFRSF9), forming a chimeric ... More
H2O2-induced endothelial NO production contributes to vascular cell apoptosis and increased permeability in rat venules.
Authors:Zhou X, Yuan D, Wang M, He P,
Journal:Am J Physiol Heart Circ Physiol
PubMed ID:23086988
Although elevated levels of H(2)O(2) have been implicated to play important roles in the pathogenesis of various cardiovascular diseases, the underlying mechanisms remain unclear. This study aims to examine the effect of H(2)O(2) on endothelial nitric oxide (NO) production in intact venules, and elucidate the role and mechanisms of NO ... More
Programmed reduction of ABC transporter activity in sea urchin germline progenitors.
Authors:Campanale JP, Hamdoun A,
Journal:Development
PubMed ID:22274698
ATP-binding cassette (ABC) transporters protect embryos and stem cells from mutagens and pump morphogens that control cell fate and migration. In this study, we measured dynamics of ABC transporter activity during formation of sea urchin embryonic cells necessary for the production of gametes, termed the small micromeres. Unexpectedly, we found ... More
Connexin-43 upregulation in micrometastases and tumor vasculature and its role in tumor cell attachment to pulmonary endothelium.
Authors:Elzarrad MK, Haroon A, Willecke K, Dobrowolski R, Gillespie MN, Al-Mehdi AB,
Journal:BMC Med
PubMed ID:18647409
The modulation of gap junctional communication between tumor cells and between tumor and vascular endothelial cells during tumorigenesis and metastasis is complex. The notion of a role for loss of gap junctional intercellular communication in tumorigenesis and metastasis has been controversial. While some of the stages of tumorigenesis and metastasis, ... More
The cellular mechanisms of neuronal swelling underlying cytotoxic edema.
Authors:Rungta RL, Choi HB, Tyson JR, Malik A, Dissing-Olesen L, Lin PJ, Cain SM, Cullis PR, Snutch TP, MacVicar BA,
Journal:
PubMed ID:25910210
Cytotoxic brain edema triggered by neuronal swelling is the chief cause of mortality following brain trauma and cerebral infarct. Using fluorescence lifetime imaging to analyze contributions of intracellular ionic changes in brain slices, we find that intense Na(+) entry triggers a secondary increase in intracellular Cl(-) that is required for ... More