CyQUANT™ NF Cell Proliferation Assay, 200 assays kit - FAQs
CyQUANT™ NF Cell Proliferation Assay, 200 assays kit - FAQs
View additional product information for CyQUANT™ Cell Proliferation Assays - FAQs (C35012, C35013, C35011, C7027, C7026, C35006, C35007)
2 product FAQs found
What is the difference between the original CyQuant assay and the CyQuant NF assay?
The original CyQUANT assay provides sensitive detection of cells over a 1000-fold linear dynamic range. In this assay, a freeze-thaw cell lysis step is required to facilitate the interaction of the CyQUANT GR dye with DNA. The CyQUANT NF assay avoids this freeze-thaw step by using a DNA binding dye in combination with a plasma membrane permeabilization reagent. The CyQUANT NF protocol requires only aspiration of growth medium (for adherent cells), replacement with dye binding solution, incubation for 30-60 minutes, and then measurement of fluorescence in a microplate reader. The assay is designed to produce a linear analytical response from at least 100-20,000 cells per well in most cell lines in a 96-well microplate.
Do you offer any products to measure neuronal cell health?
PrestoBlue Cell Viability Stain and CyQUANT Cell Proliferation Assay Kit can be used. We also offer a Neurite Outgrowth Staining Kit (Cat. No. A15001). More information about our different assays for neuronal cell health can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/neuroscience/neuronal-cell-health-assays.html).