CyQUANT™ NF Direct Red Cell Proliferation Assay - FAQs
CyQUANT™ NF Direct Red Cell Proliferation Assay - FAQs
View additional product information for CyQUANT™ Cell Proliferation Assays - FAQs (C35014)
3 product FAQs found
Can I use CyQUANT Direct Red Cell Proliferation Assay on bacteria, fungi, or other non-mammalian cell types?
We have not tested CyQUANT Direct Red Cell Proliferation Assay on prokaryotes, fungi, or any other non-mammalian cell types.
Are the components of the CyQUANT Direct Red Cell Proliferation Assay and CyQuant Direct Cell Proliferation Assay kits interchangeable?
No. The components of the CyQUANT Direct Red Cell Proliferation Assay and CyQuant Direct Cell Proliferation Assay kits are not interchangeable.
What is the difference between the CyQUANT Direct Red Cell Proliferation Assay (Cat. Nos. C35013, C35014) and the green fluorescent CyQuant Direct Cell Proliferation Assay (Cat Nos. C35011, C35012)?
The two kits utilize different dyes and background suppressors, but they function in the same way and provide similar results as can be seen from Figures 1 and 2 in the CyQUANT Direct Red Cell Proliferation Assay product sheet (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018644_CyQUANT_direct_red_assay_PI.pdf). Dyes from both kits are nucleic acid stains and the background suppressors are cell impermeant reagents that quench extracellular fluorescence. They are usable within the same range (100 to 20,000 cells/well in a 96-well format) and in the same workflow.