Carboxyl Latex Beads, 4% w/v, 3 μm - FAQs
Carboxyl Latex Beads, 4% w/v, 3 μm - FAQs
View additional product information for Carboxyl Latex Beads, 4% w/v, 3 μm - FAQs (C37281)
1 product FAQs found
Do you offer an alternative to the discontinued Carboxyl Latex Beads, 4% w/v, 3 μm (Cat. No. C37281)?
Unfortunately, we do not have a direct alternative for the discontinued Carboxyl Latex Beads, 4% w/v, 3 μm (Cat. No. C37281). However, we do offer Carboxyl Latex Beads, 4% w/v of various sizes. You can find the products using the link below.
Carboxyl Latex Beads
Additionally, Carboxyl Latex Beads, 4% w/v, 3 μm may be available as a custom order.