8mm Clear Glass Crimp Top Vial
Thermo Scientific™
8mm Clear Glass Crimp Top Vial
Select from our portfolio of high-quality glass 8mm crimp/snap top vials that are ideal low-volume sample vials for 6mm, 7mm and 8mm autosampler trays.
Have Questions?
Catalog NumberVolume (Metric)Base StyleMaterialProduct Type
C4008-741775 μLFlat BottomGlassCrimp Top Vial
Catalog number C4008-741
Price (MXN)
-
Volume (Metric):
775 μL
Base Style:
Flat Bottom
Material:
Glass
Product Type:
Crimp Top Vial
Select from our portfolio of high-quality glass 8mm crimp/snap top vials that are ideal low-volume sample vials for 6mm, 7mm and 8mm autosampler trays. Choose from flat or conical base to maximize recovery needed for small-sample injections. Re-usable sleeves support the crimped, sealed vials in the correct position in autosampler carousels or in racks and allow movement of the vials as a unit to injection positions in both GC and HPLC autosamplers. Made of superior quality borosilicate clear (Type 1, Class A), these vials meet all requirements of US, EU and JPN Pharmacopia. Choose from Thermo Scientific™ Premiere™, Thermo Scientific™ National™, Thermo Scientific™ Chromacol™ brands.
Choose the right vial for your needs
  • 8mm crimp top seal minimizes exposure between sample solvent and septum
  • Neutral Borosilicate-GOLD Grade glass is a low expansion, high purity glass with an extremely low concentration of active sites to protect your analytes from unwanted interactions
  • Precision-machined (glass or PTFE) sleeves allow the vials to be used in the vast majority of commercial autosamplers
GC and HPLC autosampler compatibility*
Thermo Scientific™
Agilent™
Beckman™
Carlo Erba™
CTC™
Fisons™
PerkinElmer™
Shimadzu™
VWR™ (Merck)/Hitachi™
*See the Thermo Scientific Columns and Consumables Catalog for autosampler compatibility
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Base StyleFlat Bottom
Closure Size8 mm
DescriptionClear Glass Vial, 7 x 40mm, Flat Bottom
Diameter (Metric) Inner7 mm
Diameter (Metric) Opening8 mm
For Use With (Equipment)Beckman™ 504, CTC/Leap™ Technologies HTS PAL, Thermo Scientific™ AS 200S, Gilson™ 231
Height (Metric)40 mm
Product TypeCrimp Top Vial
Volume (Metric) Residual<70 μL
Volume (Metric) Total775 μL
Volume (Metric) Usable650μL
Closure TypeCrimp Cap
ColorClear
Diameter (Metric) Outer7 mm
Marking SpotNo
MaterialGlass
Quantity100/Pk.
Volume (Metric)775 μL
Unit SizePack of 100