Thermo Scientific™
Aluminum Crimp Seals with Prefitted Septa for 12 x 32mm Vials
 Related applications:
Industrial Chromatography
You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Anodized aluminum seals fit all 11mm crimp-top vials
  • Seal 2000™ material is specially formulated for improved background performance over PTFE/red rubber
  • Viton™ septa perform well with ECD detectors and are resistant to many aggressive solvents
Documents
Manuals & protocols