Thermo Scientific™
8mm Autosampler Vial Septa
Catalog number: C4013-40
Thermo Scientific™
8mm Autosampler Vial Septa
Catalog number: C4013-40
Thermo Scientific™ Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of HPLC, GC and IC applications. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Catalog Number
C4013-40
Unit Size
Pack of 100
Description
Septum for 8-425 Screw Caps
Material
PTFE
Price (USD)
36.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionSeptum for 8-425 Screw Caps
ColorRed
MaterialPTFE
Pre-slitYes
SeptumRed PTFE/White Silicone/Red PTFE Septum
Material (Septa)PTFE/Silicone
Hardness45 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE, Silicone
Quantity100
Thickness (Metric)1.0 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)8 to 425 Screw Cap
Unit SizePack of 100
Catalog Number
Unit Size
Case of 1000
Description
Septum for 8-425 Screw Caps
Material
PTFE
Price (USD)
50.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionSeptum for 8-425 Screw Caps
ColorWhite
MaterialPTFE
Pre-slitYes
SeptumWhite Virgin PTFE, 0.01 in. Septµm
Material (Septa)PTFE
Hardness53 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE
Quantity1000
Thickness (Metric)0.25 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)8 to 425 Screw Cap
Unit SizeCase of 1000
Catalog Number
Unit Size
Pack of 100
Description
Septum for 8-425 Screw Caps
Material
PTFE
Price (USD)
18.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionSeptum for 8-425 Screw Caps
ColorIvory
MaterialPTFE
Pre-slitYes
Material (Septa)PTFE/Rubber
Hardness45 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE, Rubber
Quantity100
Thickness (Metric)1.0 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425mm
For Use With (Equipment)8 to 425 Screw Cap
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
Septum for 8-425 Screw Caps
Material
PTFE
Price (USD)
45.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionSeptum for 8-425 Screw Caps
ColorBlue
MaterialPTFE
Pre-slitYes
SeptumBlue PTFE/White Silicone, Pre-slit Septum
Material (Septa)PTFE/Silicone
Hardness55 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE, Silicone
Quantity100
Thickness (Metric)0.9 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)13 to 425 Screw Cap
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
Septum for 8-425 Screw Caps
Material
PTFE
Price (USD)
18.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionSeptum for 8-425 Screw Caps
ColorRed
MaterialPTFE
Pre-slitYes
SeptumRed PTFE/White Silicone Septum
Material (Septa)PTFE/Silicone
Hardness45 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE, Silicone
Quantity100
Thickness (Metric)1.3 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)8 to 425 Screw Cap
Unit SizePack of 100
Catalog Number
Unit Size
Case of 1000
Description
Septum for 8-425 Screw Caps
Material
PTFE
Price (USD)
227.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionSeptum for 8-425 Screw Caps
ColorWhite
MaterialPTFE
Pre-slitYes
Material (Septa)PTFE/Silicone
Hardness45 °shore
Product LineKimble™
Closure MaterialPTFE, Rubber
Quantity1000
Thickness (Metric)1.5 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)8 to 425 Screw Cap
Unit SizeCase of 1000
Catalog Number
Unit Size
Each
Description
Septum for 8-425 Screw Caps
Material
PTFE
Price (USD)
Availability
***
Full specifications
Quantity
-
DescriptionSeptum for 8-425 Screw Caps
ColorBlue
MaterialPTFE
Pre-slitYes
Material (Septa)PTFE/Silicone
Hardness20 °shore
Product LineAutosampler Vial Septa
Quantity500
Thickness (Metric)1.4 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8 mm
For Use With (Equipment)8 to 425 Screw Cap
Unit SizeEach
Catalog Number
Unit Size
-
Description
8mm Seal Silicone/PTFE
Material
PTFE
Price (USD)
Availability
***
Full specifications
Quantity
-
Description8mm Seal Silicone/PTFE
ColorBlue
MaterialPTFE
Pre-slitYes
Material (Septa)PTFE/Silicone
Hardness30 °shore
Product LineAutosampler Vial Septa
Quantity500
Thickness (Metric)1.1 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
For Use With (Equipment)8 to 425 Screw Thread Cap
Unit Size-
Catalog Number
Unit Size
Each
Description
Septum for 8-425 Screw Caps
Material
PTFE
Price (USD)
Availability
***
Full specifications
Quantity
-
DescriptionSeptum for 8-425 Screw Caps
ColorClear
MaterialPTFE
Pre-slitYes
SeptumRubber/PTFE
Material (Septa)PTFE/Rubber
Hardness38 °shore
Product LineAutosampler Vial Septa
Quantity1000
Thickness (Metric)1.0 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
For Use With (Equipment)8 to 425 Screw Thread Cap
Unit SizeEach
Catalog Number
Unit Size
-
Description
Septum for 8-425 Screw Caps
Material
PTFE
Price (USD)
Availability
***
Full specifications
Quantity
-
DescriptionSeptum for 8-425 Screw Caps
ColorRed
MaterialPTFE
Pre-slitYes
SeptumSilicone/PTFE
Material (Septa)PTFE/Silicone
Hardness45 °shore
Product LineAutosampler Vial Septa
Quantity500
Thickness (Metric)1.3 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
For Use With (Equipment)8 to 425 Screw Thread Cap
Unit Size-
Showing 10 of 15Show more
Catalog NumberSpecificationsUnit SizeDescriptionMaterialPrice (USD)AvailabilityQuantity
C4013-40Full specifications
Pack of 100Septum for 8-425 Screw CapsPTFE
36.00
Your Price:
***
DescriptionSeptum for 8-425 Screw Caps
ColorRed
MaterialPTFE
Pre-slitYes
SeptumRed PTFE/White Silicone/Red PTFE Septum
Material (Septa)PTFE/Silicone
Hardness45 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE, Silicone
Quantity100
Thickness (Metric)1.0 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)8 to 425 Screw Cap
Unit SizePack of 100
C4013-10Full specifications
Case of 1000Septum for 8-425 Screw CapsPTFE
50.00
Your Price:
***
DescriptionSeptum for 8-425 Screw Caps
ColorWhite
MaterialPTFE
Pre-slitYes
SeptumWhite Virgin PTFE, 0.01 in. Septµm
Material (Septa)PTFE
Hardness53 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE
Quantity1000
Thickness (Metric)0.25 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)8 to 425 Screw Cap
Unit SizeCase of 1000
C4013-30Full specifications
Pack of 100Septum for 8-425 Screw CapsPTFE
18.00
Your Price:
***
DescriptionSeptum for 8-425 Screw Caps
ColorIvory
MaterialPTFE
Pre-slitYes
Material (Septa)PTFE/Rubber
Hardness45 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE, Rubber
Quantity100
Thickness (Metric)1.0 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425mm
For Use With (Equipment)8 to 425 Screw Cap
Unit SizePack of 100
C4013-32Full specifications
Pack of 100Septum for 8-425 Screw CapsPTFE
45.00
Your Price:
***
DescriptionSeptum for 8-425 Screw Caps
ColorBlue
MaterialPTFE
Pre-slitYes
SeptumBlue PTFE/White Silicone, Pre-slit Septum
Material (Septa)PTFE/Silicone
Hardness55 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE, Silicone
Quantity100
Thickness (Metric)0.9 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)13 to 425 Screw Cap
Unit SizePack of 100
C4013-60Full specifications
Pack of 100Septum for 8-425 Screw CapsPTFE
18.00
Your Price:
***
DescriptionSeptum for 8-425 Screw Caps
ColorRed
MaterialPTFE
Pre-slitYes
SeptumRed PTFE/White Silicone Septum
Material (Septa)PTFE/Silicone
Hardness45 °shore
Product LineAutosampler Vial Septa
Closure MaterialPTFE, Silicone
Quantity100
Thickness (Metric)1.3 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)8 to 425 Screw Cap
Unit SizePack of 100
C4013-61Full specifications
Case of 1000Septum for 8-425 Screw CapsPTFE
227.00
Your Price:
***
DescriptionSeptum for 8-425 Screw Caps
ColorWhite
MaterialPTFE
Pre-slitYes
Material (Septa)PTFE/Silicone
Hardness45 °shore
Product LineKimble™
Closure MaterialPTFE, Rubber
Quantity1000
Thickness (Metric)1.5 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8-425
For Use With (Equipment)8 to 425 Screw Cap
Unit SizeCase of 1000
8-ST144Full specifications
EachSeptum for 8-425 Screw CapsPTFERequest A Quote***-
DescriptionSeptum for 8-425 Screw Caps
ColorBlue
MaterialPTFE
Pre-slitYes
Material (Septa)PTFE/Silicone
Hardness20 °shore
Product LineAutosampler Vial Septa
Quantity500
Thickness (Metric)1.4 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size8 mm
For Use With (Equipment)8 to 425 Screw Cap
Unit SizeEach
60180-515Full specifications
-8mm Seal Silicone/PTFEPTFERequest A Quote***-
Description8mm Seal Silicone/PTFE
ColorBlue
MaterialPTFE
Pre-slitYes
Material (Septa)PTFE/Silicone
Hardness30 °shore
Product LineAutosampler Vial Septa
Quantity500
Thickness (Metric)1.1 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
For Use With (Equipment)8 to 425 Screw Thread Cap
Unit Size-
8-6RT1Full specifications
EachSeptum for 8-425 Screw CapsPTFERequest A Quote***-
DescriptionSeptum for 8-425 Screw Caps
ColorClear
MaterialPTFE
Pre-slitYes
SeptumRubber/PTFE
Material (Septa)PTFE/Rubber
Hardness38 °shore
Product LineAutosampler Vial Septa
Quantity1000
Thickness (Metric)1.0 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
For Use With (Equipment)8 to 425 Screw Thread Cap
Unit SizeEach
8-ST15Full specifications
-Septum for 8-425 Screw CapsPTFERequest A Quote***-
DescriptionSeptum for 8-425 Screw Caps
ColorRed
MaterialPTFE
Pre-slitYes
SeptumSilicone/PTFE
Material (Septa)PTFE/Silicone
Hardness45 °shore
Product LineAutosampler Vial Septa
Quantity500
Thickness (Metric)1.3 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
For Use With (Equipment)8 to 425 Screw Thread Cap
Unit Size-
Showing 10 of 15Show more
  • PTFE Disk septa are chemically inert
  • PFTE red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin gauge needles
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.