13mm Autosampler Vial Screw Thread Caps
Thermo Scientific™

13mm Autosampler Vial Screw Thread Caps

13mm Screw Thread Caps feature a 13-425 thread finish. Available in an assortment of styles, pre-assembled caps and septa minimize contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSeptumMaterialProduct Type
C4015-55BLKBlack, Blue, WhitePTFE/Silicone, Pre-slitPolypropylene, PTFE, SiliconeOpen Top Screw Cap
C4015-1ABlackNonePolypropyleneOpen Top Screw Cap
C4015-66ABlack, Red, WhiteRed PTFE/White SiliconePhenolic, PTFE, SiliconeOpen Top Screw Cap
C4015-30ABlack, Ivory, RedIvory PTFE/Red RubberPolypropylene, PTFE, RubberOpen Top Screw Cap
C4015-40ABlack, Red, WhitePTFE/Silicone/PTFEPolypropylene, PTFE, SiliconeOpen Top Screw Cap
C4015-5ANaturalIntegral MoldedPolypropylene, PolyethyleneScrew Cap
12-SCR
also known as 12-SC(R)
RedNonePolypropyleneOpen Top Screw Cap
12-SCY
also known as 12-SC(Y)
YellowNonePolypropyleneOpen Top Screw Cap
Catalog number C4015-55BLK
Price (BRL)
-
Color:
Black, Blue, White
Septum:
PTFE/Silicone, Pre-slit
Material:
Polypropylene, PTFE, Silicone
Product Type:
Open Top Screw Cap
Thermo Scientific™ 13mm Screw Thread Caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs. Convenient pre-assembled caps and septa minimize contamination from handling. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Autosampler Vial Screw Thread Caps
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure Size13 mm
Color (Seal)Blue/White
DescriptionPP, Black; Blue PTFE/White Silicone Preslit septum
For Use With (Equipment)13-425 Screw Thread Vials
Product TypeOpen Top Screw Cap
SeptumPTFE/Silicone, Pre-slit
Thickness (Metric)1.5 mm
ColorBlack, Blue, White
MaterialPolypropylene, PTFE, Silicone
Pre-slitYes
Quantity100
Unit SizePack of 100