Thermo Scientific™
9mm Assembled Clear Autosampler Vial Kits
Catalog number: C5000-186
Thermo Scientific™
9mm Assembled Clear Autosampler Vial Kits
Catalog number: C5000-186
Save time during sample preparation and reduce the risk of contamination using Thermo Scientific™ 9mm Clear Vial Convenience Kits. Each kit includes 100 clear glass vials pre-assembled with 100 screw thread caps and septa. They are packaged in convenient vial trays with clear covers or in economical polybags. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Catalog Number
C5000-186
Unit Size
Pack of 100
Septum
Red PTFE/White Silicone/Red PTFE
Marking Spot
No
Color
Clear
Material
Glass
Price (USD)
74.00
Your Price:
Full specifications
Quantity
Closure ColorBlue
DescriptionAssembled AVCS Kit
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumRed PTFE/White Silicone/Red PTFE
IncludesVial: C4000-1, Closure: C5000-53B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotNo
Septa Size (Metric)9 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Septum
Ivory PTFE/Red Rubber
Marking Spot
No
Color
Clear
Material
Glass
Price (USD)
56.00
Your Price:
Full specifications
Quantity
Closure ColorBlue
DescriptionClear Glass; Ivory PTFE/Red Rubber Septum; Blue DP Cap
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumIvory PTFE/Red Rubber
IncludesVial: C4000-1, Closure: C5000-51B
Material (Septa)PTFE/Rubber
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotNo
Septa Size (Metric)9 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Septum
Ivory PTFE/Red Rubber
Marking Spot
Yes
Color
Clear
Material
Glass
Price (USD)
58.00
Your Price:
Full specifications
Quantity
Closure ColorBlue
DescriptionClear Glass w/I-D Patch, Ivory PTFE/RR Septum, Blue DP Cap
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumIvory PTFE/Red Rubber
IncludesVial: C4000-1W, Closure: C5000-51B
Material (Septa)PTFE/Rubber
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Septum
Ivory PTFE/Red Rubber
Marking Spot
Yes
Color
Clear
Material
Glass
Price (USD)
74.00
Your Price:
Full specifications
Quantity
Closure ColorBlue
DescriptionAssembled AVCS Kit
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitYes
SeptumIvory PTFE/Red Rubber
IncludesVial: C4000-1W, Closure: C5000-57B
Material (Septa)PTFE/Rubber
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Septum
Red PTFE/White Silicone/Red PTFE
Marking Spot
Yes
Color
Clear
Material
Glass
Price (USD)
73.00
Your Price:
Full specifications
Quantity
Closure ColorBlue
DescriptionClear Glass w/I-D Patch, Red PTFE/White Silicone/Red PTFE septum, Blue DP Cap
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumRed PTFE/White Silicone/Red PTFE
IncludesVial: C4000-1W, Closure: C5000-53B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Septum
Red PTFE/White Silicone
Marking Spot
No
Color
Clear
Material
Glass
Price (USD)
63.00
Your Price:
Full specifications
Quantity
Closure ColorBlue
DescriptionAssembled AVCS Kit
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumRed PTFE/White Silicone
IncludesVial: C4000-1, Closure: C5000-54B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotNo
Septa Size (Metric)9 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Septum
Red PTFE/White Silicone
Marking Spot
Yes
Color
Clear
Material
Glass
Price (USD)
64.00
Your Price:
Full specifications
Quantity
Closure ColorBlue
DescriptionClear Glass w/I-D Patch, Red PTFE/White Silicone Septum, Blue DP Cap
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumRed PTFE/White Silicone
IncludesVial: C4000-1W, Closure: C5000-54B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Septum
Blue PTFE/White Silicone
Marking Spot
Yes
Color
Clear
Material
Glass
Price (USD)
83.00
Your Price:
Full specifications
Quantity
Closure ColorBlue
DescriptionAssembled AVCS Kit
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitYes
SeptumBlue PTFE/White Silicone
IncludesVial: C4000-1W, Closure: C5000-55B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeSeptumMarking SpotColorMaterialPrice (USD)Quantity
C5000-186Full specifications
Pack of 100Red PTFE/White Silicone/Red PTFENoClearGlass
74.00
Your Price:
Closure ColorBlue
DescriptionAssembled AVCS Kit
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumRed PTFE/White Silicone/Red PTFE
IncludesVial: C4000-1, Closure: C5000-53B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotNo
Septa Size (Metric)9 mm
Unit SizePack of 100
C5000-180Full specifications
Pack of 100Ivory PTFE/Red RubberNoClearGlass
56.00
Your Price:
Closure ColorBlue
DescriptionClear Glass; Ivory PTFE/Red Rubber Septum; Blue DP Cap
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumIvory PTFE/Red Rubber
IncludesVial: C4000-1, Closure: C5000-51B
Material (Septa)PTFE/Rubber
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotNo
Septa Size (Metric)9 mm
Unit SizePack of 100
C5000-180WFull specifications
Pack of 100Ivory PTFE/Red RubberYesClearGlass
58.00
Your Price:
Closure ColorBlue
DescriptionClear Glass w/I-D Patch, Ivory PTFE/RR Septum, Blue DP Cap
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumIvory PTFE/Red Rubber
IncludesVial: C4000-1W, Closure: C5000-51B
Material (Septa)PTFE/Rubber
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
C5000-183WFull specifications
Pack of 100Ivory PTFE/Red RubberYesClearGlass
74.00
Your Price:
Closure ColorBlue
DescriptionAssembled AVCS Kit
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitYes
SeptumIvory PTFE/Red Rubber
IncludesVial: C4000-1W, Closure: C5000-57B
Material (Septa)PTFE/Rubber
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
C5000-186WFull specifications
Pack of 100Red PTFE/White Silicone/Red PTFEYesClearGlass
73.00
Your Price:
Closure ColorBlue
DescriptionClear Glass w/I-D Patch, Red PTFE/White Silicone/Red PTFE septum, Blue DP Cap
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumRed PTFE/White Silicone/Red PTFE
IncludesVial: C4000-1W, Closure: C5000-53B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
C5000-192Full specifications
Pack of 100Red PTFE/White SiliconeNoClearGlass
63.00
Your Price:
Closure ColorBlue
DescriptionAssembled AVCS Kit
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumRed PTFE/White Silicone
IncludesVial: C4000-1, Closure: C5000-54B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotNo
Septa Size (Metric)9 mm
Unit SizePack of 100
C5000-192WFull specifications
Pack of 100Red PTFE/White SiliconeYesClearGlass
64.00
Your Price:
Closure ColorBlue
DescriptionClear Glass w/I-D Patch, Red PTFE/White Silicone Septum, Blue DP Cap
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitNo
SeptumRed PTFE/White Silicone
IncludesVial: C4000-1W, Closure: C5000-54B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
C5000-195WFull specifications
Pack of 100Blue PTFE/White SiliconeYesClearGlass
83.00
Your Price:
Closure ColorBlue
DescriptionAssembled AVCS Kit
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass
Pre-slitYes
SeptumBlue PTFE/White Silicone
IncludesVial: C4000-1W, Closure: C5000-55B
Material (Septa)PTFE/Silicone
No. per Pack100
Product LineSurestop
Volume (Metric) Total2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open Top, 6 mm hole)
Height (Metric)32 mm
TypeAssembled Screw Vial AVCS Convenience Kit
Closure Size9 mm
Marking SpotYes
Septa Size (Metric)9 mm
Unit SizePack of 100
Showing 8 of 8
Features of Target DP 9mm Vial Closures (included in kits):
  • Septa push through nearly eliminated due to improved interior geometry without the need for bonding
  • Increased sealing capability
  • AVCS provides the freedom to select the best septum for your instrument and applications
  • Cost efficient alternative to current choices for resolving push through (bonded septa)
  • Optimized ergonomics, fine texturing and evenly spaced ribbing for superior handling even with poorly fitting gloves
  • Rough edges have been eliminated making it easier and more comfortable to use while improving autosampler operation
  • National C5000 Series Target DP 9mm Wide Opening 9mm Closure
    Utilizing AVCS technology, the C5000 series wide opening closure increases the available injection area by over 7% providing greater area for injection needle penetration.
    National C5000 Series Target DP 9mm Integral Membrane Closure
    In addition to AVCS technology, the C5000 series closure provides a halogen and rubber-free sample handling environment for HPLC applications or applications where silicones and rubber septa are not suitable.
    National C5000 Series Target DP 9mm Solid Closure
    AVCS technology provides maximum protection of samples during long term storage. The protective top prevents accidental puncture of the septum and resulting sample loss. Can be stored down to -80°F.
    Ordering Information:
    All products in quantity of 100/PK.
    Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.