Notice: This product will be discontinued on September 1, 2017. For superior performance, we recommend the TOP10F´ Electrocomp™ Kit.

TOP10F´ Electrocomp™ Kit

Catalog number: C66524

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The TOP10F' genotype is identical to TOP10 with the addition of an F' episome. The F' episome carries the tetracycline resistance gene and allows isolation of single-stranded DNA from vectors that have an f1 origin of replication. In addition, the F' carries the lacIq repressor for inducible expression from trc, tac, and lac promoters using IPTG. TOP10F' require IPTG induction for blue/white color screening.

The TOP10F' genotype offers the following features:

hsdR for efficient transformation of unmethylated DNA from PCR amplifications
mcrA for efficient transformation of methylated DNA from genomic preparations
lacZΔM15 for blue/white color screening of recombinant clones
endA1 for cleaner preparations of DNA and better results in downstream applications due to the elimination of non-specific digestion by Endonuclease I
recA1 for reduced occurrence of non-specific recombination in cloned DNA
lacIq repressor for inducible expression from trc, tac, and lac promoters

Note: This kit contains enough electrocompetent cells for 75 x 40 µL rxns or 150 x 20 µL rxns. Please see the product manual for reaction scale recommendations.

Genotype:
F'{lacIqTn10(TetR)} mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK rpsL endA1 nupG
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Competent Cell Type: Electrocompetent
Species: E. coli
Format: Tube(s)
Transformation Efficiency: > 1x10^9
Bacterial or Yeast Strain: TOP10F´
Product Line: Electrocomp™
Blue-White Screening: Yes
Cloning Methylated DNA: Yes
Cloning Unstable DNA: Not Suitable for cloning unstable DNA
F' Episome (to make ssDNA): Contains F' Episome
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Improves Plasmid Quality: Yes
Plasmid Type: May be used for plasmids > 20 kb
Preparing Unmethylated DNA: Not Suitable for preparing unmethylated DNA
Product Size: 30 x 100 µL
Propagating ccdB Vectors: Not for ccdB vector propagation
Reduces Recombination: Yes
T1 Phage - Resistant (tonA): No

Documents

Manuals & protocols

Recommended products