Cell Culture Contamination Detection Kit - FAQs
Cell Culture Contamination Detection Kit - FAQs
View additional product information for Cell Culture Contamination Detection Kit - FAQs (C7028)
2 product FAQs found
What does the Cell Culture Contamination Detection kit screen for?
This kit can screen tissue cultures for contamination by microorganisms. The kit not only serves to detect the contaminants, it also identifies the contaminant type. Three fluorescent dyes distinctly stain yeast (and other fungi), gram-positive and gram-negative bacteria in slide preparations. The kit provides results within one hour, and, by staining the microorganisms with different fluorescent colors, it helps a scientist choose an appropriate course of action.
Can the Cell Culture Contamination Detection Kit (Cat. No. C7028) detect mycoplasma?
Yes, the Cell Culture Contamination Detection Kit will detect/stain mycoplasma. The SYTO-9 cell-permeant dye will stain every cell, including the mammalian cells (if present). Mycoplasma will be seen as small contaminating cells in the medium. It may be challenging to identify Mycoplasma from other possible contaminating species, if present.
We also offer the MycoFluor Mycoplasma Detection Kit (Cat. No. M7006) for mycoplasma detection in lab cultures.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.