Product Image
Thermo Scientific™
AquaSensors DataStick CPVC Communications Adaptor
AquaSensors DataStick CPVC Comms Adaptor Ethernet IP Modbus TCP and direct PC and PLC applications with RJ45 connector for PoE 9-metre cable for use with DataStick body
Have Questions?
Número de catálogo CA27R3A
Precio (MXN)
-
AquaSensors DataStick CPVC Comms Adaptor Ethernet IP Modbus TCP and direct PC and PLC applications with RJ45 connector for PoE 9-metre cable for use with DataStick body
Communications adapter
Plugs into the DataStick body to provide power and direct interactive communications with control systems
Especificaciones
Temperatura de funcionamiento (métrico)-20°C to +65°C
TipoCommunications Adapter
Unit SizeEach