Can I use StemPro HSC Expansion Medium (Prototype) without cytokines or growth factors?
This media system is formulated without cytokines or growth factors, but cytokines and/or growth factors should be added immediately prior to use. For hematopoietic stem cell culture, we recommend adding the following cytokines at the final concentration indicated: SCF (100 ng/mL), FLT-3L (100 ng/mL), TPO (100 ng/mL), IL-3 (50 ng/mL), and IL-6 (20 ng/mL).