COUNT-CAL™ Count Precision Size Standards, 15μm, 15.02μm ± 0.15μm, σ = 0.15μm, 1.0% CV - FAQs
COUNT-CAL™ Count Precision Size Standards, 15μm, 15.02μm ± 0.15μm, σ = 0.15μm, 1.0% CV - FAQs
View additional product information for COUNT-CAL™ Count Precision Size Standards - FAQs (CC05-PK, CC30-PK, CC02-PK, CC15-PK, CC10-PK, CC20-PK, CC25-PK, CC50-PK, CC70-PK)
1 product FAQs found
How should I store COUNT-CAL Count Precision Size Standards?
Unless otherwise stated, we recommend refrigerating COUNT-CAL Count Precision Size Standards at 2 to 8 degrees C when not in use. We do not recommenf freezing them. The bottle must be kept tightly sealed and stored upright.