Nalgene™ PETG Ultra-Low Particulate Media Bottles - FAQs
Nalgene™ PETG Ultra-Low Particulate Media Bottles - FAQs
View additional product information for Nalgene™ PETG Ultra-Low Particulate Media Bottles - FAQs (CE-N2019-0060, CE-N2019-0125, CE-N2019-0500, CE-N2019-0030, CE-N2019-0250, CE-N2019-1000, CE-N2019-2000)
1 product FAQs found
What is the difference between the following Nalgene Square PETG Media Bottles: Cat. No. 2019-xxxx (with Closure), Cat. No. 342020-xxxx (with Closure: Sterile, Shrink-Wrapped Trays), Cat. No. CE-N2019-xxxx (Platinum Certified Clean)?
All three products use the same PETG bottles, so the materials and specifications are the same for the bottle itself. Cat. No. 342020-xxxx is supplied with natural-colored HDPE closures, while Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx are supplied with white HDPE closures. The HDPE resin used for the closures is the same, but a white colorant has been added to the resin for Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx (this will be noted on the COA). Cat. No. 2019-xxxx also has a PVC tamper-resistant seal around the closure, while Cat. No. 342020-xxxx does not.
For more detail about the different "levels of clean" for our Thermo Scientific Nalgene contains, please refer to our Nalgene Clean Containers brochure.