Nalgene™ PETG Ultra-Low Particulate Media Bottles
Thermo Scientific™

Nalgene™ PETG Ultra-Low Particulate Media Bottles

Designed for the most critical applications, Nalgene PETG Ultra-Low Particulate Square Media Bottles are platinum certified to contain less than one-third of the USP <788> particulate limits. With exceptionally low particulates and endotoxins, they set the standard for biological and pharmaceutical manufacturing downstream applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
CE-N2019-0250250 mL
CE-N2019-003030 mL
CE-N2019-006060 mL
CE-N2019-0125125 mL
CE-N2019-0500500 mL
CE-N2019-10001 L
CE-N2019-20002 L
Catalog number CE-N2019-0250
Price (INR)
-
Capacity (Metric):
250 mL

Products are ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates. They are also well suited for the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Eliminates costly washing, depyrogenation and autoclaving steps
  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates
  • Also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
  • Supported by extensive validation documentation
Specifications
AutoclavableNot Autoclavable
Capacity (English)8 oz.
Certifications/ComplianceUSP Class VI Compliant, Certified Radiation Stabilized (SAL 10-6)
Closure Size38-430 mm
Diameter (English)2.4 in.
Diameter (English) Inner Neck1.10 in.
Diameter (Metric)59 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Height (English)5.75 in.
Height (Metric) With Closure144 mm
Low Particulate<1/3 USP <788> (Ultralow particulate)
No. per Case48/Cs.
No. per Pack12/Pk.
Product TypeMedia Bottle
QuantityCase of 48
Temperature Range (Metric)-70°C to 70°C
Weight (English)10.2 lb.
Width (English)12.12 in.
Workflow StepDownstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Capacity (Metric)250 mL
ColorClear
MaterialPETG
MouthNarrow
SterilitySterile
Unit SizeCase of 48

Frequently asked questions (FAQs)

What is the difference between the following Nalgene Square PETG Media Bottles: Cat. No. 2019-xxxx (with Closure), Cat. No. 342020-xxxx (with Closure: Sterile, Shrink-Wrapped Trays), Cat. No. CE-N2019-xxxx (Platinum Certified Clean)?

All three products use the same PETG bottles, so the materials and specifications are the same for the bottle itself. Cat. No. 342020-xxxx is supplied with natural-colored HDPE closures, while Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx are supplied with white HDPE closures. The HDPE resin used for the closures is the same, but a white colorant has been added to the resin for Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx (this will be noted on the COA). Cat. No. 2019-xxxx also has a PVC tamper-resistant seal around the closure, while Cat. No. 342020-xxxx does not.

For more detail about the different "levels of clean" for our Thermo Scientific Nalgene contains, please refer to our Nalgene Clean Containers brochure.