Nalgene™ PETG Ultra-Low Particulate Media Bottles
Thermo Scientific™

Nalgene™ PETG Ultra-Low Particulate Media Bottles

Designed for the most critical applications, Nalgene PETG Ultra-Low Particulate Square Media Bottles are platinum certified to contain less than one-third of the USP <788> particulate limits. With exceptionally low particulates and endotoxins, they set the standard for biological and pharmaceutical manufacturing downstream applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
CE-N2019-0500500 mL
CE-N2019-003030 mL
CE-N2019-006060 mL
CE-N2019-0125125 mL
CE-N2019-0250250 mL
CE-N2019-10001 L
CE-N2019-20002 L
Catalog number CE-N2019-0500
Price (BRL)
-
Capacity (Metric):
500 mL

Products are ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates. They are also well suited for the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Eliminates costly washing, depyrogenation and autoclaving steps
  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates
  • Also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
  • Supported by extensive validation documentation
Specifications
AutoclavableNot Autoclavable
Closure Size38-430 mm
Diameter (English)2.9 in.
Diameter (English) Inner Neck1.10 in.
Diameter (Metric)74 mm
Diameter (Metric) Inner Neck28 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Height (English)6.97 in.
Height (Metric) With Closure177 mm
Low Particulate<1/3 USP <788> (Ultralow particulate)
No. per Case70/Cs.
No. per Pack5/Pk.
Temperature Range (Metric)-70°C to 70°C
Weight (English)7.6 lb.
Width (English)15.6 in.
Workflow StepDownstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Capacity (English)16 oz.
Capacity (Metric)500 mL
Certifications/ComplianceUSP Class VI Compliant, Certified Radiation Stabilized (SAL 10-6)
ColorClear
MaterialPETG
MouthNarrow
Product TypeMedia Bottle
SterilitySterile
Unit SizeCase of 70