Frasco de PETG cuadrado para medios de cultivo con certificación de limpieza óptima Nalgene™
Thermo Scientific™

Frasco de PETG cuadrado para medios de cultivo con certificación de limpieza óptima Nalgene™

Designed for the most critical applications, Nalgene PETG Ultra-Low Particulate Vials are platinum certified to contain less than one-third of the USP <788> particulate limits. With exceptionally low particulates and endotoxins, they set the standard for biological and pharmaceutical manufacturing downstream applications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)
CE-N2035-002020 ml
CE-N2035-00055 ml
Número de catálogo CE-N2035-0020
Precio (CLP)
-
Capacidad (métrico):
20 ml

Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Sterile to 10-6 SAL
  • Non-pyrogenic and non-cytotoxic
  • Leakproof High Density Polyethylene (HDPE) closures
  • Serum Vials are packed in shrink-wrapped trays with closures attached
  • Manufactured in compliance with ISO 13485:2003 quality requirements
Especificaciones
Certificaciones/conformidadISO 13485:2003, 21CFR177.1520, Clase VI de la Farmacopea Estadounidense (USP)
Para utilizar con (aplicación)Muestreo, almacenamiento y envío estériles, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Niveles bajos de partículas<1/3 USP <788> (Ultralow particulate)
N.º por caja504/Cs.
N.° por paquete84/Pk.
Tipo de productoMedia Bottle
CantidadCase of 504
Intervalo de temperatura (métrico)-70°C to 70°C
Paso del flujo de trabajoDownstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Capacidad (métrico)20 ml
EsterilidadEstéril
Unit SizeCase of 504