Specifications
AutoclavableNot Autoclavable
Capacity (English)0.67 oz.
Certifications/ComplianceUSP Class VI
Closure LinerLined
Closure MaterialPolypropylene
Closure Size20 mm
Closure TypeCap
Diameter (Metric)30 mm
For Use With (Application)Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product
Height (English)2.72 in.
Height (Metric)69 mm
Liner MaterialSilicone
Product TypeStorage and Transport Bottle
ShapeRound
Temperature Range (Metric)-100°C to 100°C
Through the Closure DispensingNo
Width (English)11 in.
Workflow StepDownstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Capacity (Metric)20 mL
Closure IncludedYes
ColorClear
GraduatedYes
MaterialPolycarbonate
MouthNarrow
SterilitySterile
Unit SizeEach