DataTaker Analog Channel Expansion Module
Thermo Fisher Scientific™
DataTaker Analog Channel Expansion Module
20-60 Analog Channel Expansion Module for DataTaker
Have Questions?
Catalog NumberChannels
CEM2020 to 60 Analog
Catalog number CEM20
Price (USD)
-
Channels:
20 to 60 Analog
Increasing the channel capacity of the DataTaker DT80 range is made very easy by adding DataTaker Channel Expansion Modules (CEM20). Each CEM20 connects 20 universal data logging channels to the DataTaker data logger.
Increasing the channel capacity of the DataTaker DT80 range is made very easy by adding DataTaker Channel Expansion Modules (CEM20). Each CEM20 connects 20 universal data logging channels to the DataTaker data logger.
DataTaker enclosures give you the flexibility to install your DataTaker system in a harsh or otherwise unsuitable environment by providing protection against external conditions such as water, dust, and accidental damage. These enclosures are suitable for all dataTaker systems and can be used for, industrial, weather proof or portable applications.
Compatible with DT80 & DT85 series loggers. (Series 2 and later)
Specifications
CalibrationNot Calibrated
Channels20 to 60 Analog
DescriptionAnalog Channel Expansion Module - 20-60 Analog Channels
Display TypeNo Display
TypeAnalog Channel Expansion Module
Unit SizeEach