SAA Human Matched Antibody Pair - FAQs
SAA Human Matched Antibody Pair - FAQs
View additional product information for SAA Human Matched Antibody Pair - FAQs (CHA2513)
1 product FAQs found
Does the SAA Human ELISA Kit (Cat. No. KHA0011) use the same antibodies as the Human SAA Antibody Pair (Cat. No. CHA2513)?
Yes, these kits use the same antibodies.