Human IL-8 Matched Antibody Pair - FAQs
Human IL-8 Matched Antibody Pair - FAQs
View additional product information for Human IL-8 Matched Antibody Pair - FAQs (CHC1303)
1 product FAQs found
Are the antibodies in IL-8 Human Matched Antibody Pair (Cat. No. CHC1303) both monoclonal?
Yes, both the capture antibody and the detection antibody in IL-8 Human Matched Antibody Pair (Cat. No. CHC1303) are monoclonal.