Do Human IL-12/IL-23 p40 ELISA Kit (Cat. No. KHC0231) and Human IL-12/IL-23 p40 Human Matched Antibody Pair (Cat. No. CHC2493) use the same antibodies?
The kits use the same capture antibodies, but the detection antibodies are different.